VWR® Talon® Extra-Large Lab-Frames | Avantor
VWR® Talon® Extra-Large Lab-Frames
Supplier:   VWR International
VWR® Talon® Extra-Large Lab-Frames
Supplier:  VWR International
Certificates
VWR® Talon® Extra-Large Lab-Frames
VWR® Talon® Extra-Large Lab-Frames
2 Options Available Below
Product Details & Documents
Extra-large lab frames offer the greatest versatility for your most demanding setups.
  • Extra large design
  • For horizontal or vertical use
  • Versatile and adaptable
  • Mountable on either side
Measure 1220Lx1830Hmm (48x72"). May be used either horizontally or vertically. Include six 305mm (12") rods and feet for securing frame to wall. Apparatus mountable on either side of frame.
Ordering information: Each Lab Frame includes six 51 mm (2") rods, six 305 mm (12") rods, two 610 mm (24") rods, seven 1220 mm (48") rods, five 1829 mm (72") rods, four rod end connectors, forty-five S-connectors, and eight Lab Frame feet.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Availability
    Price
  • Lab-Frame with Aluminum Rods
    Aluminum
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,557.10
    $4,557.10
    Prix promotionnel pour les nouveaux clients
    Each
       $4,557.10
  • Lab-Frame with Stainless Steel Rods
    Stainless steel
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,090.12
    $6,090.12
    Each
       $6,090.12