VWR® Spectrosil Fluorometer Cells
À propos de cet article
Standard rectangular cells manufactured from Spectrosil quartz for accurate readings over the entire UV range to near infrared range.
- Far UV quartz
- Wavelength range of 170–2700 nm
- Fully heat-fused construction
Due to the fully heat-fused construction (no intermediate materials or adhesives) and careful annealing to remove any strain, there is no optical or physical distortion of the four optically polished windows
Lids and stoppers are made of polytetrafluoroethylene (PTFE). Cell 414004-057 features a closed-top polymer screw cap with a silicone rubber gasket. Cell 414004-058 features an open-top polymer screw cap with a silicone rubber septum.
Cells exhibit a transmission tolerance within 1.5% at 200nm.
Ordering information: For Spectrosil cells for use in spectrophotometers, see 414004-060 series.
4 Options Available Below
Product Details & Documents
Standard rectangular cells manufactured from Spectrosil quartz for accurate readings over the entire UV range to near infrared range.
- Far UV quartz
- Wavelength range of 170–2700 nm
- Fully heat-fused construction
Due to the fully heat-fused construction (no intermediate materials or adhesives) and careful annealing to remove any strain, there is no optical or physical distortion of the four optically polished windows
Lids and stoppers are made of polytetrafluoroethylene (PTFE). Cell 414004-057 features a closed-top polymer screw cap with a silicone rubber gasket. Cell 414004-058 features an open-top polymer screw cap with a silicone rubber septum.
Cells exhibit a transmission tolerance within 1.5% at 200nm.
Ordering information: For Spectrosil cells for use in spectrophotometers, see 414004-060 series.
Product Family Options
Informations sur le produit
- Internal DimensionsSample VolumePathlengthCover TypeExterior DimensionsAvailabilityPrice
- W×L 10×10 mm3.5 ml10 mmStopper12.5W x 12.5L x 45H mmLog in pour la disponibilitéPack of 1Pack of 1 - $540.34$540.34Prix promotionnel pour les nouveaux clientsLog in pour la disponibilitéCase of 8Case of 8 - $4,322.69$4,322.69Prix promotionnel pour les nouveaux clientsPack of 1$540.34Case of 8$4,322.69
- W×L 10×10 mm3.5 ml10 mmFlat lid12.5W x 12.5L x 45H mmLog in pour la disponibilitéPack of 2Pack of 2 - $1,864.87$1,864.87Prix promotionnel pour les nouveaux clientsLog in pour la disponibilitéCase of 16Case of 16 - $14,918.98$14,918.98Prix promotionnel pour les nouveaux clientsPack of 2$1,864.87Case of 16$14,918.98
- W×L 10×10 mm3.5 ml10 mmScrew cap12.5W x 12.5L x 52H mmLog in pour la disponibilitéPack of 1Pack of 1 - $558.19$558.19Prix promotionnel pour les nouveaux clientsLog in pour la disponibilitéCase of 8Case of 8 - $4,465.51$4,465.51Prix promotionnel pour les nouveaux clientsPack of 1$558.19Case of 8$4,465.51
- W×L 10×10 mm3.5 ml10 mmSeptum screw cap12.5W x 12.5L x 52H mmLog in pour la disponibilitéPack of 1Pack of 1 - $777.82$777.82Prix promotionnel pour les nouveaux clientsLog in pour la disponibilitéCase of 8Case of 8 - $6,222.59$6,222.59Prix promotionnel pour les nouveaux clientsPack of 1$777.82Case of 8$6,222.59