VWR® Talon® Large Lab-Frames | Avantor
VWR® Talon® Large Lab-Frames
Supplier:   VWR International
VWR® Talon® Large Lab-Frames
Supplier:  VWR International
Certificates
VWR® Talon® Large Lab-Frames
VWR® Talon® Large Lab-Frames
2 Options Available Below
Product Details & Documents
These 1220x1220mm (48x48") frames are ideal for complex glassware setups.
  • For large-capacity setups
  • Stable
  • Ideal for complex glass setups
Base is 460mm (18") wide for stability.
Ordering information: Each Lab Frame includes ten 51 mm (2") rods, three 457 mm (18") rods, two 914 mm (36") rods, ten 1220 mm (48") rods, four rod end connectors, thirty-eight S-connectors, and six Lab Frame feet.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Availability
    Price
  • Lab-Frame with Aluminum Rods
    Aluminum
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,582.92
    $3,582.92
    Prix promotionnel pour les nouveaux clients
    Each
       $3,582.92
  • Lab-Frame with Stainless Steel Rods
    Stainless steel
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,095.22
    $4,095.22
    Prix promotionnel pour les nouveaux clients
    Each
       $4,095.22