VWR® Talon® Medium Lab-Frames | Avantor
VWR® Talon® Medium Lab-Frames
Supplier:   VWR International
VWR® Talon® Medium Lab-Frames
Supplier:  VWR International
Certificates
VWR® Talon® Medium Lab-Frames
VWR® Talon® Medium Lab-Frames
2 Options Available Below
Product Details & Documents
Lab Frames are convenient for distillation and general setups.
  • For horizontal or vertical mounting
  • Convenient size for distillation and general setups
  • Stable base
Can be assembled as 1220Lx610Hmm (48x24") horizontal or 610Lx1220Hmm (24x48") vertical frame. Base is 46 cm (18") wide for stability.
Ordering information: Each Lab Frame includes eight 51 mm (2") rods, two 457 mm (18") rods, seven 610 mm (24") rods, four 1220 mm (48") rods, four rod end connectors, thirty-five S-connectors, and four Lab Frame feet.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Availability
    Price
  • Lab-Frame with Aluminum Rods
    Aluminum
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,156.10
    $2,156.10
    Prix promotionnel pour les nouveaux clients
    Each
       $2,156.10
  • Lab-Frame with Stainless Steel Rods
    Stainless steel
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,592.16
    $2,592.16
    Prix promotionnel pour les nouveaux clients
    Each
       $2,592.16