White Plastic Screw Caps, WHEATON®
White plastic screw caps fit Wheaton vials (see 66021-679 series) and other glass scintillation vials.
- Suitable for marking
- Not autoclavable
- Available unlined or choose from a variety of liners
White plastic screw caps fit Wheaton vials (see 66021-679 series) and other glass scintillation vials.
- Suitable for marking
- Not autoclavable
- Available unlined or choose from a variety of liners
Product Family Options
- Taille du bouchonCap TypeLiner Material
- : 240805
- : 241017
- : 240917
- : 240804
- : 241317
- : 241009
- : 241018
- : 240817
- : 240818