VWR® Maximum Protection Fluid-Impervious Ankle-High Boot Covers
VWR® Maximum Protection Fluid-Impervious Ankle-High Boot Covers
VWR® Maximum Protection Fluid-Impervious Ankle-High Boot Covers
VWR® Maximum Protection Fluid-Impervious Ankle-High Boot Covers
VWR® Maximum Protection Fluid-Impervious Ankle-High Boot Covers
These boot covers are the cleanest and most durable within the entire VWR® Protection line of apparel.
  • Fluid-impervious material with seamless bottom to repel water
  • Skid resistance on wet and smooth surfaces
  • Excellent particulate barrier
  • Low linting
Engineered to optimize barrier protection, cleanliness, and comfort
Boot covers are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validated through independent lab testing.
Appropriate for industrial, pharmaceutical, food processing, and certain environmental cleanup applications. Serged seams.
Documents
  • Catalog No:
  • 414004-638
  • 414004-639
  • 414004-640
  • Garment application:
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Bacterial filtration efficiency:
  • 0.999
  • 0.999
  • 0.999
  • Particle shedding (Helmke Drum):
  • Level I
  • Level I
  • Level I
  • Water resistance:
  • Pass
  • Pass
  • Pass
  • Tensile strength:
  • 28.8 (Filling)
  • 28.8 (Filling)
  • 28.8 (Filling)
  • Tearing strength:
  • 16.5 (Widthwise Yarns)
  • 16.5 (Widthwise Yarns)
  • 16.5 (Widthwise Yarns)
  • Weight:
  • 60.21 g/m²
  • 60.21 g/m²
  • 60.21 g/m²
Product Family Options
  • Color
    Pointure
    Hauteur
    Longueur
  • Blue
    XL
    28 cm (11")
    42 cm (16½")
    Case of 200
    Case of 200 - $347.79
    $347.79
    Pallet of 3600
    Pallet of 3600 - $6,260.26
    $6,260.26
    Case of 200
       $347.79
    Pallet of 3600
       $6,260.26
    Specifications
    Cat. No.
    414004-638
    Garment application
    Cleanroom
    Bacterial filtration efficiency
    0.999
    Particle shedding (Helmke Drum)
    Level I
    Water resistance
    Pass
    Tensile strength
    28.8 (Filling)
    Tearing strength
    16.5 (Widthwise Yarns)
    Weight
    60.21 g/m²
  • Black
    XXL
    28.6 cm (11¼")
    47 cm (18½")
    Case of 200
    Case of 200 - $356.64
    $356.64
    Pallet of 3600
    Pallet of 3600 - $6,419.52
    $6,419.52
    Case of 200
       $356.64
    Pallet of 3600
       $6,419.52
    Specifications
    Cat. No.
    414004-639
    Garment application
    Cleanroom
    Bacterial filtration efficiency
    0.999
    Particle shedding (Helmke Drum)
    Level I
    Water resistance
    Pass
    Tensile strength
    28.8 (Filling)
    Tearing strength
    16.5 (Widthwise Yarns)
    Weight
    60.21 g/m²
  • White
    Universal
    28 cm (11")
    38.1 cm (15")
    Case of 200
    Case of 200 - $337.65
    $337.65
    Pallet of 3600
    Pallet of 3600 - $6,077.66
    $6,077.66
    Case of 200
       $337.65
    Pallet of 3600
       $6,077.66
    Specifications
    Cat. No.
    414004-640
    Garment application
    Cleanroom
    Bacterial filtration efficiency
    0.999
    Particle shedding (Helmke Drum)
    Level I
    Water resistance
    Pass
    Tensile strength
    28.8 (Filling)
    Tearing strength
    16.5 (Widthwise Yarns)
    Weight
    60.21 g/m²