Ordinary Chondrite
Ordinary chondrite, <1 g wt.
Catalog # 470355-906
Supplier: AEROLITE METEORITES INC SE
: A-ST-BOX
Jump to:
Product Details & Documents
Economical and compact meteorite collectible.
- 100% guaranteed to be authentic and as described
- Acrylic presenetation box
- Information card included
Stones constitute the largest group of meteorites. They originated in the outer crust of a planet or asteroid. Visible inside most stone meteorites are tiny, glassy, spheres known as chondrules. Forged at the very dawn of the solar system, these chondrules are far older than our own planet. Some stone meteorites, known as carbonaceous chondrites, have been found to contain water, salt, and even amino acids. In the distant past, these meteorites may have carried the very building blocks of life to earth.
Because no two meteorites or impactities are the same, yours will be similar but not exact.
: Package is 2×1.5".
Spécifications
- Désignation:Ordinary chondrite
- Taille:<1 g wt.