VWR® Glass Pocket Thermometers
Fournisseur: VWR International
Certificats
À propos de cet article
Glass pocket thermometers are conveniently sized and accurate to within one degree.
- Red liquid-filled
- Fahrenheit scale
Thermometers have permanently fused markings. Accuracy: ±1 division. Length: 160mm (65/16"). Each includes a heavy-duty case.
Frequently Bought Together