ContactUsLinkComponent
SP Bel-Art Mini Magnifier, Bel-Art Products, a part of SP
SP Bel-Art Mini Magnifier, Bel-Art Products, a part of SP
Catalog # 54848-306
undefined
SP Bel-Art Mini Magnifier, Bel-Art Products, a part of SP
Catalog # 54848-306
Supplier Number:  F37865-0000
Specifications
  • Magnification:
    1.75×
  • Cat. no.:
    54848-306
  • W×D:
    14.3×10.5 cm (5⅝×4⅛")
  • Supplier No.:
    378650000
Specifications
About this item
Freestanding magnifier permits enhanced viewing of slides, Petri dishers, multiwell plates, gels, or text.
  • Freestanding design
  • Can be placed over a variety of objects
  • Simple to use
Precision-molded lens offers 1.75 magnification. The unique stand allows the magnifier to be placed over a variety of objects.
Stand height: 17.2 cm (63/4")
Unit weight: 368.5 g (13 oz.)