Biohazard Wipes and Liners, Current Technologies
Biohazard Wipes and Liners, Current Technologies
Biohazard Wipes and Liners, Current Technologies
Certificates
Biohazard Wipes and Liners, Current Technologies
Biohazard Wipes and Liners, Current Technologies
12 Options Available Below
Product Details & Documents
Low-linting wipes and liners are specifically designed to protect workers and labs from contamination from blood, body fluids, tissue, specimens plus chemicals.
Wipes and liners are available in several sizes and in large rolls for cutting to size. Wipes are also available in standard absorbency, which can absorb 9× its own weight, heavy absorbency, which can absorb 12× its weight and super absorbency, which can absorb 25× its weight.
Specifically sized dispensers are constructed of clear acrylic.
Absorbent white side soaks up blood, tissue, specimen, body fluids, and chemicals. Bright orange barrier side, made of coextruded plastic, blocks soak-through to gloves, hands and counters, limiting the spread of contamination. Wipes are impermeable to strong solvents including xylene and xylene substitutes, acetone, ketones, alcohols, and formalin. Use to pop caps, wipe pipets and probes, handle specimens and line benches, counters and trays, clean up spills.
Ordering information: Bio-Screen wipes and liners are non-sterile and are not for use on patients.
Product Family Options
Informations sur le produit
  • Désignation
    Dimensions
    Availability
    Price
  • Standard Absorbency Biohazard Wipes
    7.6 x 7.6 cm (3 x 3")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 2,000
    Case of 2,000 - $586.65
    $586.65
    Case of 2,000
       $586.65
    Supplier Number: BH33400
  • Standard Absorbency Biohazard Wipes
    10.2 x 10.2 cm (4 x 4")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 2,000
    Case of 2,000 - $692.71
    $692.71
    Case of 2,000
       $692.71
    Supplier Number: BH44985
  • Standard Absorbency Biohazard Wipes, Roll
    40.6 cm x 30.5 m (16" x 100')
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 2
    Case of 2 - $768.03
    $768.03
    Case of 2
       $768.03
    Supplier Number: BH16100
  • Heavy Absorbency Biohazard Wipes
    7.6 x 7.6 cm (3 x 3")
    Discontinued
    Supplier Number: BH32000
  • Heavy Absorbency Biohazard Wipes
    10.2 x 10.2 cm (4 x 4")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 2,000
    Case of 2,000 - $402.63
    $402.63
    Case of 2,000
       $402.63
    Supplier Number: BH44200
  • Heavy Absorbency Biohazard Wipes
    27.9 x 40.6 cm (11 x 16")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 200
    Case of 200 - $431.81
    $431.81
    Case of 200
       $431.81
    Supplier Number: BH11016ET
  • Heavy Absorbency Biohazard Wipes
    40.6 x 40.6 cm (16 x 16")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 2
    Case of 2 - $565.58
    $565.58
    Case of 2
       $565.58
    Supplier Number: BH 10600
  • Heavy Absorbency Biohazard Wipes
    40.6 x 40.6 cm (16 x 16")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 125
    Case of 125 - $501.18
    $501.18
    Case of 125
       $501.18
    Supplier Number: BH 16016 ET
  • Super Absorbency Biohazard Wipes
    7.6 x 7.6 cm (3 x 3")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,800
    Case of 1,800 - $603.73
    $603.73
    Case of 1,800
       $603.73
    Supplier Number: BH91033
  • Super Absorbency Biohazard Wipes
    10.2 x 10.2 cm (4 x 4")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,800
    Case of 1,800 - $602.01
    $602.01
    Case of 1,800
       $602.01
    Supplier Number: BH90044
  • Super Absorbency Biohazard Wipes
    20.3 x 22.9 cm (8 x 9")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 360
    Case of 360 - $611.40
    $611.40
    Case of 360
       $611.40
    Supplier Number: BH92910
  • Super Absorbency Biohazard Wipes
    40.6 x 40.6 cm (16 x 16")
    Discontinued
    Supplier Number: -
  • Super Absorbency Biohazard Wipes
    40.6 x 63.5 cm (16 x 25")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 60
    Case of 60 - $561.07
    $561.07
    Case of 60
       $561.07
    Supplier Number: BH91625
  • Super Absorbency Biohazard Wipes, Roll
    40.6 cm x 30.5 m (16" x 100')
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 2
    Case of 2 - $889.43
    $889.43
    Case of 2
       $889.43
    Supplier Number: BH93016
Accessories
Informations sur le produit
  • Catalog #
    Material
    Availability
    Price
  • Dispenser for 10.2 x 10.2 cm (4 x 4") Wipes/Towels
    Dispenser for 10.2 x 10.2 cm (4 x 4") Wipes/Towels
    16200-180
    Acrylic
    Log in pour la disponibilité
    Each - $97.22
Informations sur le produit
  • Catalog #
    Material
    Availability
    Price
  • Dispenser for 7.6 x 7.6 cm (3 x 3") Wipes/Towels
    Dispenser for 7.6 x 7.6 cm (3 x 3") Wipes/Towels
    16003-778
    Acrylic
    Log in pour la disponibilité
    Each - $97.65