Synthware Liebig Condensers
Supplier: Kemtech America
Jump to:
À propos de cet article
With top outer [ST] joint and bottom inner [ST] joint.
- Liebig style condenser
- Heavy wall borosilicate construction
- Ground glass joint
- Jacketed
Jacketed liebig condenser is perfect for most chemical distillation applications.
9 Options Available Below
Product Details & Documents
With top outer [ST] joint and bottom inner [ST] joint.
- Liebig style condenser
- Heavy wall borosilicate construction
- Ground glass joint
- Jacketed
Jacketed liebig condenser is perfect for most chemical distillation applications.
Product Family Options
8 mm Hose Connection
Informations sur le produit
10 mm Hose Connection
Informations sur le produit
- ST JointJacket LengthAvailabilityPrice
- Supplier Number: C204100
- Supplier Number: C204200
- Supplier Number: C204250
- Supplier Number: C204300
- Supplier Number: C204400
- Supplier Number: C204500