VWR® Storage Racks for 80-Well Microtube Racks
Racks allow organization of large number of microtubes easily on benchtops.
- Sturdy
- Acrylic construction
- Accommodate up to eight 80-well microtube racks
Racks accommodate up to eight 80-well microtube racks with either 1.5 mL or 2.0 mL microtubes in the racks.
Frequently Bought Together