ContactUsLinkComponent
SP Bel-Art Microcentrifuge Tube Opener, Bel-Art Products, a part of SP
Catalogue # 47743-758
undefined
SP Bel-Art Microcentrifuge Tube Opener, Bel-Art Products, a part of SP
Catalogue # 47743-758
Supplier Number:  F19925-0001
Spécifications
  • Description:
    Microcentrifuge Tube Opener, 79 mm (31/8")
  • Cat. no.:
    47743-758
  • Supplier No.:
    199250001
Spécifications
À propos de cet article
This convenient tool for opening microcentrifuge tubes is double-ended to work on all tube sizes.
  • Double-ended design
  • Open milli- and microliter-sized tubes
  • Convenient design and handling
One end opens standard 1.5, 2.0, and 2.2 mL tubes while the other end has a forked blade for opening smaller sized tubes — 250, 400, and 550µL.