InterGuard™ Sleeve, InterWorld™
White, 45.7 cm (18")
Catalog # 37002-062
Supplier: Interworld
Supplier #: 10135-102-02
Jump to:
Product Details & Documents
Sleeve is designed to provide barrier protection in dry applications
It offers a secure fit with elastic at both ends. Constructed of lightweight, breathable, spunbonded polypropylene.
Ordering information: Packaged 100 sleeves per bag, 3 bags per case.
Spécifications
- Color:White
- Longueur:45.7 cm (18")
- Cat. No.:37002-062
- Garment application:Laboratory