Corning® Cap, Phenolic, PTFE Liner
 :   Corning
Corning® Cap, Phenolic, PTFE Liner
 :  Corning
Corning® Cap, Phenolic, PTFE Liner
Corning® Cap, Phenolic, PTFE Liner
This cap is fabricated of a resin specially formulated to resist the effects of temperature and steam in autoclaving.
  • Available in different sizes
Caps are provided with a glued-in rubber liner over which is firmly bonded a PTFE interface which provides a highly inert sealing face.
  • Catalog No:
  • 60827-225
  • 60827-227
  • 60827-228
  • 60827-229
  • 66110-220
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
Product Family Options
  • Hauteur
    Thread Finish
    Color
    Taille du bouchon
  • 12,5 mm
    24
    Black
    24-400
    Case of 12
    Case of 12 - 95,70 $
    95,70 $
    Case of 12
       95,70 $
     : 9998-24
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    60827-225
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 13,5 mm
    13
    Black
    13-415
    Case of 288
    Case of 288 - 905,56 $
    905,56 $
    Case of 288
       905,56 $
     : 9998-13
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    60827-227
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 16 mm
    15
    Black
    15-415
    Case of 288
    Case of 288 - 841,75 $
    841,75 $
    Case of 288
       841,75 $
     : 9998-15
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    60827-228
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 17,5 mm
    18
    Black
    18-415
    Case of 192
    Case of 192 - 411,09 $
    411,09 $
    Case of 192
       411,09 $
     : 9998-18
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    60827-229
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 18,5 mm
    Black
    24-410
    Case of 144
    Case of 144 - 725,00 $
    725,00 $
    Case of 144
       725,00 $
     : 9998-241
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    66110-220
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact