VWR® Basic SMS Coveralls
VWR® Basic SMS Coveralls
Supplier:  VWR International
Certificates
VWR® Basic SMS Coveralls
35 Options Available Below
Product Details & Documents
VWR® Basic Protection SMS Coveralls are manufactured from a latex-free, multi-layer spunbonded-meltblown-spunbonded (SMS) fabric that offers high tensile strength while remaining soft, comfortable, and breathable. These static dissipative garments act as a fluid and particulate barrier and provide light chemical splash resistance.
  • SMS (Spunbond-Meltblown-Spunbond) nonwoven fabric
  • Multi-layer protection combines strength, filtration, and comfort
  • Outer spunbond layers allow airflow while maintaining barrier integrity
  • Middle meltblown layer provides enhanced protection against particulates and limited liquid splashes
  • Zipper front closure, elastic wrists and ankles
  • Tear-resistant and abrasion-resistant for extended wear
  • Effective barrier against dust, dry particulates, and light liquid splashes
  • Breathable design reduces heat stress during long shifts
  • Meets industry standards for nonhazardous protective apparel
The SMS coverall is engineered for environments requiring enhanced protection without sacrificing comfort. Constructed from SMS fabric (Spunbond-Meltblown-Spunbond), this material combines three layers to deliver superior performance. The outer spunbond layers provide strength and durability, while the meltblown middle layer acts as an effective barrier against particulates and limited liquid splashes. This multi-layer design ensures a balance of breathability, protection, and flexibility, making SMS coveralls ideal for extended wear in controlled environments.
Key features include a zipper front closure for secure fit, and elastic wrists and ankles to minimize exposure gaps. SMS coveralls are non-linting, reducing contamination risks, and are widely used in pharmaceutical, cleanroom, food processing, and industrial settings where particulate control and splash resistance are critical.
Unlike basic polypropylene garments, SMS coveralls offer higher barrier protection while maintaining comfort, making them suitable for tasks involving powders, dry particulates, and light liquid contact. Their disposable nature supports hygiene compliance and simplifies inventory management, providing a practical and cost-effective solution for facilities prioritizing both safety and efficiency.
 : Coveralls are packaged 25 per case.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 76045-406
  • 76318-834
  • 76318-836
  • 76318-838
  • 76319-110
  • 76320-012
  • 76320-014
  • 76320-016
  • 76320-076
  • 76320-078
  • 414004-333
  • 414004-692
  • 414004-693
  • 414004-694
  • 414004-331
  • 414004-330
  • 414004-329
  • 414004-334
  • 414004-332
  • 89234-870
  • 89234-872
  • 414004-328
  • 414004-327
  • 414004-325
  • 414004-324
  • 414004-322
  • 414004-323
  • 414004-326
  • 89234-874
  • 89234-876
  • 414004-687
  • 414004-688
  • 414004-689
  • 414004-690
  • 414004-691
  • Fabric:
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Closure type:
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Cleanroom class:
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • Pockets:
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Sleeve or leg length:
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Garment application:
  • Cleanroom|Industrial|Laboratory|General Purpose
  • Cleanroom|Industrial|Laboratory|General Purpose
  • Cleanroom|Industrial|Laboratory|General Purpose
  • Cleanroom|Industrial|Laboratory|General Purpose
  • Cleanroom|Industrial|Laboratory|General Purpose
  • Cleanroom|Industrial|Laboratory|General Purpose
  • Cleanroom|Industrial|Laboratory|General Purpose
  • Cleanroom|Industrial|Laboratory|General Purpose
  • Cleanroom|Industrial|Laboratory|General Purpose
  • Cleanroom|Industrial|Laboratory|General Purpose
  • Cleanroom|Industrial|Laboratory|General Purpose
  • Cleanroom|Industrial|Laboratory|General Purpose
  • Cleanroom|Industrial|Laboratory|General Purpose
  • Cleanroom|Industrial|Laboratory|General Purpose
  • Cleanroom|Industrial|Laboratory|General Purpose
  • Cleanroom|Industrial|Laboratory|General Purpose
  • Cleanroom|Industrial|Laboratory|General Purpose
  • Cleanroom|Industrial|Laboratory|General Purpose
  • Cleanroom|Industrial|Laboratory|General Purpose
  • Cleanroom|Industrial|Laboratory|General Purpose
  • Cleanroom|Industrial|Laboratory|General Purpose
  • Cleanroom|Industrial|Laboratory|General Purpose
  • Cleanroom|Industrial|Laboratory|General Purpose
  • Cleanroom|Industrial|Laboratory|General Purpose
  • Cleanroom|Industrial|Laboratory|General Purpose
  • Cleanroom|Industrial|Laboratory|General Purpose
  • Cleanroom|Industrial|Laboratory|General Purpose
  • Cleanroom|Industrial|Laboratory|General Purpose
  • Cleanroom|Industrial|Laboratory|General Purpose
  • Cleanroom|Industrial|Laboratory|General Purpose
  • Cleanroom|Industrial|Laboratory|General Purpose
  • Cleanroom|Industrial|Laboratory|General Purpose
  • Cleanroom|Industrial|Laboratory|General Purpose
  • Cleanroom|Industrial|Laboratory|General Purpose
  • Cleanroom|Industrial|Laboratory|General Purpose
  • Air permeability:
  • 0.9 cu. ft./min./sq. ft.
  • 0.9 cu. ft./min./sq. ft.
  • 0.9 cu. ft./min./sq. ft.
  • 0.9 cu. ft./min./sq. ft.
  • 0.9 cu. ft./min./sq. ft.
  • 0.9 cu. ft./min./sq. ft.
  • 0.9 cu. ft./min./sq. ft.
  • 0.9 cu. ft./min./sq. ft.
  • 0.9 cu. ft./min./sq. ft.
  • 0.9 cu. ft./min./sq. ft.
  • 0.9 cu. ft./min./sq. ft.
  • 0.9 cu. ft./min./sq. ft.
  • 0.9 cu. ft./min./sq. ft.
  • 0.9 cu. ft./min./sq. ft.
  • 0.9 cu. ft./min./sq. ft.
  • 0.9 cu. ft./min./sq. ft.
  • 0.9 cu. ft./min./sq. ft.
  • 0.9 cu. ft./min./sq. ft.
  • 0.9 cu. ft./min./sq. ft.
  • 0.9 cu. ft./min./sq. ft.
  • 0.9 cu. ft./min./sq. ft.
  • 95.3 cu. ft./min./sq. ft.
  • 0.9 cu. ft./min./sq. ft.
  • 0.9 cu. ft./min./sq. ft.
  • 0.9 cu. ft./min./sq. ft.
  • 0.9 cu. ft./min./sq. ft.
  • 0.9 cu. ft./min./sq. ft.
  • 0.9 cu. ft./min./sq. ft.
  • 0.9 cu. ft./min./sq. ft.
  • 0.9 cu. ft./min./sq. ft.
  • 0.9 cu. ft./min./sq. ft.
  • 0.9 cu. ft./min./sq. ft.
  • 0.9 cu. ft./min./sq. ft.
  • 0.9 cu. ft./min./sq. ft.
  • 0.9 cu. ft./min./sq. ft.
  • Bacterial filtration efficiency:
  • 95.30%
  • 95.30%
  • 95.30%
  • 95.30%
  • 95.30%
  • 95.30%
  • 95.30%
  • 95.30%
  • 95.30%
  • 95.30%
  • 95.30%
  • 95.30%
  • 95.30%
  • 95.30%
  • 95.30%
  • 95.30%
  • 95.30%
  • 95.30%
  • 95.30%
  • 95.30%
  • 95.30%
  • >90%
  • 95.30%
  • 95.30%
  • 95.30%
  • 95.30%
  • 95.30%
  • 95.30%
  • 95.30%
  • 95.30%
  • 95.30%
  • 95.30%
  • 95.30%
  • 95.30%
  • 95.30%
  • Particle shedding (Helmke Drum):
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Water resistance:
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • Water vapor transmission rate:
  • 965 g/m²/24 hrs.
  • 965 g/m²/24 hrs.
  • 965 g/m²/24 hrs.
  • 965 g/m²/24 hrs.
  • 965 g/m²/24 hrs.
  • 965 g/m²/24 hrs.
  • 965 g/m²/24 hrs.
  • 965 g/m²/24 hrs.
  • 965 g/m²/24 hrs.
  • 965 g/m²/24 hrs.
  • 965 g/m²/24 hrs.
  • 965 g/m²/24 hrs.
  • 965 g/m²/24 hrs.
  • 965 g/m²/24 hrs.
  • 965 g/m²/24 hrs.
  • 965 g/m²/24 hrs.
  • 965 g/m²/24 hrs.
  • 965 g/m²/24 hrs.
  • 965 g/m²/24 hrs.
  • 965 g/m²/24 hrs.
  • 965 g/m²/24 hrs.
  • 965 g/m²/24 hrs.
  • 965 g/m²/24 hrs.
  • 965 g/m²/24 hrs.
  • 965 g/m²/24 hrs.
  • 965 g/m²/24 hrs.
  • 965 g/m²/24 hrs.
  • 965 g/m²/24 hrs.
  • 965 g/m²/24 hrs.
  • 965 g/m²/24 hrs.
  • 965 g/m²/24 hrs.
  • 965 g/m²/24 hrs.
  • 965 g/m²/24 hrs.
  • 965 g/m²/24 hrs.
  • 965 g/m²/24 hrs.
  • Tensile strength:
  • 13 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13 lbs./in. (warp), 16.1 lbs./in. (filling)
  • Tearing strength:
  • 3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
  • 3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
  • 3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
  • 3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
  • 3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
  • 3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
  • 3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
  • 3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
  • 3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
  • 3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
  • 3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
  • 3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
  • 3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
  • 3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
  • 3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
  • 3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
  • 3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
  • 3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
  • 3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
  • 3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
  • 3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
  • 3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
  • 3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
  • 3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
  • 3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
  • 3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
  • 3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
  • 3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
  • 3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
  • 3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
  • 3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
  • 3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
  • 3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
  • 3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
  • 3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
  • Weight:
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
  • 50 g/m²
Spécifications
Product Family Options
Standard coveralls
Informations sur le produit
  • Désignation
    Color
    Taille de confection
    Longueur
    Chest Size
    Sleeve
    Availability
    Price
  • White
    S
    36" (body), 27" (leg)
    23½"
    32¾"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $382.54
    $382.54
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $382.54
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
    Spécifications:
    More
    Specifications
    Cat. No.
    76045-406
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory|General Purpose
    Air permeability
    0.9 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    95.30%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Water vapor transmission rate
    965 g/m²/24 hrs.
    Tensile strength
    13 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
    Weight
    50 g/m²
  • Black
    XL
    40" (body), 30" (leg)
    27"
    36½"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $480.60
    $480.60
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $480.60
    Spécifications:
    More
    Specifications
    Cat. No.
    76318-834
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory|General Purpose
    Air permeability
    0.9 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    95.30%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Water vapor transmission rate
    965 g/m²/24 hrs.
    Tensile strength
    13 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
    Weight
    50 g/m²
  • Black
    XXL
    40½" (body), 31" (leg)
    29¾"
    37"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $523.24
    $523.24
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $523.24
    Spécifications:
    More
    Specifications
    Cat. No.
    76318-836
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory|General Purpose
    Air permeability
    0.9 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    95.30%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Water vapor transmission rate
    965 g/m²/24 hrs.
    Tensile strength
    13 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
    Weight
    50 g/m²
  • Black
    3XL
    43" (body), 32" (leg)
    32"
    38"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $536.23
    $536.23
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $536.23
    Spécifications:
    More
    Specifications
    Cat. No.
    76318-838
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory|General Purpose
    Air permeability
    0.9 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    95.30%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Water vapor transmission rate
    965 g/m²/24 hrs.
    Tensile strength
    13 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
    Weight
    50 g/m²
  • Black
    L
    38¾" (body), 29½" (leg)
    25"
    34"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $463.90
    $463.90
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $463.90
    Spécifications:
    More
    Specifications
    Cat. No.
    76319-110
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory|General Purpose
    Air permeability
    0.9 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    95.30%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Water vapor transmission rate
    965 g/m²/24 hrs.
    Tensile strength
    13 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
    Weight
    50 g/m²
  • Black
    S
    36" (body), 27" (leg)
    23½"
    32¾"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $449.69
    $449.69
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $449.69
    Spécifications:
    More
    Specifications
    Cat. No.
    76320-012
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory|General Purpose
    Air permeability
    0.9 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    95.30%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Water vapor transmission rate
    965 g/m²/24 hrs.
    Tensile strength
    13 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
    Weight
    50 g/m²
  • Black
    M
    37½" (body), 28½" (leg)
    24¼"
    34"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $453.06
    $453.06
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $453.06
    Spécifications:
    More
    Specifications
    Cat. No.
    76320-014
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory|General Purpose
    Air permeability
    0.9 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    95.30%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Water vapor transmission rate
    965 g/m²/24 hrs.
    Tensile strength
    13 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
    Weight
    50 g/m²
  • Black
    4XL
    43" (body), 32½" (leg)
    32½"
    41"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $538.71
    $538.71
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $538.71
    Spécifications:
    More
    Specifications
    Cat. No.
    76320-016
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory|General Purpose
    Air permeability
    0.9 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    95.30%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Water vapor transmission rate
    965 g/m²/24 hrs.
    Tensile strength
    13 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
    Weight
    50 g/m²
  • Black
    5XL
    43½" (body), 33" (leg)
    33½"
    42"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $552.40
    $552.40
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $552.40
    Spécifications:
    More
    Specifications
    Cat. No.
    76320-076
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory|General Purpose
    Air permeability
    0.9 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    95.30%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Water vapor transmission rate
    965 g/m²/24 hrs.
    Tensile strength
    13 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
    Weight
    50 g/m²
  • Black
    7XL
    45½" (body), 35" (leg)
    36"
    44"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $566.00
    $566.00
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $566.00
    Spécifications:
    More
    Specifications
    Cat. No.
    76320-078
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory|General Purpose
    Air permeability
    0.9 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    95.30%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Water vapor transmission rate
    965 g/m²/24 hrs.
    Tensile strength
    13 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
    Weight
    50 g/m²
  • White
    M
    37½" (body), 28½" (leg)
    24¼"
    34"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $396.13
    $396.13
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 450
    Pallet of 450 - $7,130.30
    $7,130.30
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $396.13
    Pallet of 450
       $7,130.30
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-333
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory|General Purpose
    Air permeability
    0.9 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    95.30%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Water vapor transmission rate
    965 g/m²/24 hrs.
    Tensile strength
    13 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
    Weight
    50 g/m²
  • White
    L
    38¾" (body), 29½" (leg)
    25"
    34"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $398.10
    $398.10
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 600
    Pallet of 600 - $9,554.50
    $9,554.50
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $398.10
    Pallet of 600
       $9,554.50
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-331
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory|General Purpose
    Air permeability
    0.9 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    95.30%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Water vapor transmission rate
    965 g/m²/24 hrs.
    Tensile strength
    13 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
    Weight
    50 g/m²
  • White
    XL
    40" (body), 30" (leg)
    27"
    36½"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $397.06
    $397.06
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 450
    Pallet of 450 - $7,147.01
    $7,147.01
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $397.06
    Pallet of 450
       $7,147.01
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-330
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory|General Purpose
    Air permeability
    0.9 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    95.30%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Water vapor transmission rate
    965 g/m²/24 hrs.
    Tensile strength
    13 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
    Weight
    50 g/m²
  • White
    XXL
    40½" (body), 31" (leg)
    29¾"
    37"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $462.60
    $462.60
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 450
    Pallet of 450 - $8,326.80
    $8,326.80
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $462.60
    Pallet of 450
       $8,326.80
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-329
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory|General Purpose
    Air permeability
    0.9 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    95.30%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Water vapor transmission rate
    965 g/m²/24 hrs.
    Tensile strength
    13 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
    Weight
    50 g/m²
  • White
    3XL
    43" (body), 32" (leg)
    32"
    38"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $476.92
    $476.92
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 450
    Pallet of 450 - $8,584.56
    $8,584.56
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $476.92
    Pallet of 450
       $8,584.56
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-334
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory|General Purpose
    Air permeability
    0.9 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    95.30%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Water vapor transmission rate
    965 g/m²/24 hrs.
    Tensile strength
    13 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
    Weight
    50 g/m²
  • White
    4XL
    43" (body), 32½" (leg)
    32½"
    41"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $478.58
    $478.58
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 600
    Pallet of 600 - $11,485.82
    $11,485.82
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $478.58
    Pallet of 600
       $11,485.82
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-332
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory|General Purpose
    Air permeability
    0.9 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    95.30%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Water vapor transmission rate
    965 g/m²/24 hrs.
    Tensile strength
    13 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
    Weight
    50 g/m²
  • White
    5XL
    43½" (body), 33" (leg)
    33½"
    42"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $506.56
    $506.56
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 525
    Pallet of 525 - $10,637.76
    $10,637.76
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $506.56
    Pallet of 525
       $10,637.76
    Spécifications:
    More
    Specifications
    Cat. No.
    89234-870
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory|General Purpose
    Air permeability
    0.9 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    95.30%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Water vapor transmission rate
    965 g/m²/24 hrs.
    Tensile strength
    13 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
    Weight
    50 g/m²
  • White
    7XL
    45½" (body), 35" (leg)
    36"
    44"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $526.75
    $526.75
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $526.75
    Spécifications:
    More
    Specifications
    Cat. No.
    89234-872
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory|General Purpose
    Air permeability
    0.9 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    95.30%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Water vapor transmission rate
    965 g/m²/24 hrs.
    Tensile strength
    13 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
    Weight
    50 g/m²
  • Blue
    S
    36" (body), 27" (leg)
    23½"
    32¾"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $395.86
    $395.86
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 600
    Pallet of 600 - $9,500.54
    $9,500.54
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $395.86
    Pallet of 600
       $9,500.54
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-328
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory|General Purpose
    Air permeability
    95.3 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    >90%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Water vapor transmission rate
    965 g/m²/24 hrs.
    Tensile strength
    13 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
    Weight
    50 g/m²
  • Blue
    M
    37½" (body), 28½" (leg)
    24¼"
    34"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $394.46
    $394.46
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 600
    Pallet of 600 - $9,466.94
    $9,466.94
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $394.46
    Pallet of 600
       $9,466.94
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-327
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory|General Purpose
    Air permeability
    0.9 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    95.30%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Water vapor transmission rate
    965 g/m²/24 hrs.
    Tensile strength
    13 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
    Weight
    50 g/m²
  • Blue
    L
    38¾" (body), 29½" (leg)
    25"
    34"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $397.06
    $397.06
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 600
    Pallet of 600 - $9,529.34
    $9,529.34
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $397.06
    Pallet of 600
       $9,529.34
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-325
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory|General Purpose
    Air permeability
    0.9 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    95.30%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Water vapor transmission rate
    965 g/m²/24 hrs.
    Tensile strength
    13 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
    Weight
    50 g/m²
  • Blue
    XL
    40" (body), 30" (leg)
    27"
    36½"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $396.26
    $396.26
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 450
    Pallet of 450 - $7,132.61
    $7,132.61
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $396.26
    Pallet of 450
       $7,132.61
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-324
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory|General Purpose
    Air permeability
    0.9 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    95.30%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Water vapor transmission rate
    965 g/m²/24 hrs.
    Tensile strength
    13 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
    Weight
    50 g/m²
  • Blue
    XXL
    40½" (body), 31" (leg)
    29¾"
    37"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $448.78
    $448.78
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 450
    Pallet of 450 - $8,078.11
    $8,078.11
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $448.78
    Pallet of 450
       $8,078.11
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-322
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory|General Purpose
    Air permeability
    0.9 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    95.30%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Water vapor transmission rate
    965 g/m²/24 hrs.
    Tensile strength
    13 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
    Weight
    50 g/m²
  • Blue
    3XL
    43" (body), 32" (leg)
    32"
    38"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $483.56
    $483.56
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 450
    Pallet of 450 - $8,704.08
    $8,704.08
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $483.56
    Pallet of 450
       $8,704.08
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-323
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory|General Purpose
    Air permeability
    0.9 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    95.30%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Water vapor transmission rate
    965 g/m²/24 hrs.
    Tensile strength
    13 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
    Weight
    50 g/m²
  • Blue
    4XL
    43" (body), 32½" (leg)
    32½"
    41"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $485.34
    $485.34
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 450
    Pallet of 450 - $8,736.19
    $8,736.19
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $485.34
    Pallet of 450
       $8,736.19
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-326
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory|General Purpose
    Air permeability
    0.9 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    95.30%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Water vapor transmission rate
    965 g/m²/24 hrs.
    Tensile strength
    13 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
    Weight
    50 g/m²
  • Blue
    5XL
    43½" (body), 33" (leg)
    33½"
    42"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $534.30
    $534.30
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 450
    Pallet of 450 - $9,617.33
    $9,617.33
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $534.30
    Pallet of 450
       $9,617.33
    Spécifications:
    More
    Specifications
    Cat. No.
    89234-874
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory|General Purpose
    Air permeability
    0.9 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    95.30%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Water vapor transmission rate
    965 g/m²/24 hrs.
    Tensile strength
    13 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
    Weight
    50 g/m²
  • Blue
    7XL
    45½" (body), 35" (leg)
    36"
    44"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $551.58
    $551.58
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 450
    Pallet of 450 - $9,928.51
    $9,928.51
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $551.58
    Pallet of 450
       $9,928.51
    Spécifications:
    More
    Specifications
    Cat. No.
    89234-876
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory|General Purpose
    Air permeability
    0.9 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    95.30%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Water vapor transmission rate
    965 g/m²/24 hrs.
    Tensile strength
    13 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
    Weight
    50 g/m²
Coveralls with attached boot
Informations sur le produit
  • Désignation
    Color
    Taille de confection
    Longueur
    Chest Size
    Sleeve
    Availability
    Price
  • Blanc
    3XL
    43" (body), 32" (leg)
    32"
    38"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $59.33
    $59.33
    Log in pour la disponibilité
    Pallet of 450
    Pallet of 450 - $1,067.62
    $1,067.62
    Case of 25
       $59.33
    Pallet of 450
       $1,067.62
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-692
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory|General Purpose
    Air permeability
    0.9 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    95.30%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Water vapor transmission rate
    965 g/m²/24 hrs.
    Tensile strength
    13 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
    Weight
    50 g/m²
  • Blanc
    4XL
    43" (body), 32½" (leg)
    32½"
    41"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $233.66
    $233.66
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $233.66
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-693
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory|General Purpose
    Air permeability
    0.9 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    95.30%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Water vapor transmission rate
    965 g/m²/24 hrs.
    Tensile strength
    13 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
    Weight
    50 g/m²
  • Blanc
    5XL
    43½" (body), 33" (leg)
    33½"
    42"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $30.15
    $30.15
    Case of 25
       $30.15
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-694
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory|General Purpose
    Air permeability
    0.9 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    95.30%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Water vapor transmission rate
    965 g/m²/24 hrs.
    Tensile strength
    13 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
    Weight
    50 g/m²
  • White
    S
    36" (body), 27" (leg)
    23½"
    32¾"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $246.86
    $246.86
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $246.86
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-687
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory|General Purpose
    Air permeability
    0.9 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    95.30%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Water vapor transmission rate
    965 g/m²/24 hrs.
    Tensile strength
    13 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
    Weight
    50 g/m²
  • White
    M
    37½" (body), 28½" (leg)
    24¼"
    34"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $359.34
    $359.34
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $359.34
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-688
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory|General Purpose
    Air permeability
    0.9 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    95.30%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Water vapor transmission rate
    965 g/m²/24 hrs.
    Tensile strength
    13 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
    Weight
    50 g/m²
  • White
    L
    38¾" (body), 29½" (leg)
    25"
    34"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $284.98
    $284.98
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 450
    Pallet of 450 - $5,129.57
    $5,129.57
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $284.98
    Pallet of 450
       $5,129.57
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-689
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory|General Purpose
    Air permeability
    0.9 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    95.30%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Water vapor transmission rate
    965 g/m²/24 hrs.
    Tensile strength
    13 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
    Weight
    50 g/m²
  • White
    XL
    40" (body), 30" (leg)
    27"
    36½"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $359.96
    $359.96
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 450
    Pallet of 450 - $6,479.28
    $6,479.28
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $359.96
    Pallet of 450
       $6,479.28
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-690
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory|General Purpose
    Air permeability
    0.9 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    95.30%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Water vapor transmission rate
    965 g/m²/24 hrs.
    Tensile strength
    13 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
    Weight
    50 g/m²
  • White
    XXL
    40½" (body), 31" (leg)
    29¾"
    37"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $381.47
    $381.47
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 450
    Pallet of 450 - $6,866.50
    $6,866.50
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $381.47
    Pallet of 450
       $6,866.50
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-691
    Fabric
    Spunbound polypropylene
    Closure type
    Zip closure
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory|General Purpose
    Air permeability
    0.9 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    95.30%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Water vapor transmission rate
    965 g/m²/24 hrs.
    Tensile strength
    13 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
    Weight
    50 g/m²