Accessories for Temperature/Humidity Data Loggers with LCD, Dickson
Accessories for Temperature/Humidity Data Loggers with LCD, Dickson
Supplier:   Dickson
Accessories for Temperature/Humidity Data Loggers with LCD, Dickson
Supplier:  Dickson
Certificates
Accessories for Temperature/Humidity Data Loggers with LCD, Dickson
Accessories for Temperature/Humidity Data Loggers with LCD, Dickson
1 Options Available Below
Product Details & Documents
Specifications for Product Family
Spécifications
  • Catalog No:
  • 12444-120
  • 14905-018
  • 14905-020
  • W×D×H:
  • Ø 150 mm (6")
  • Ø 150 mm (6")
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Range
    Availability
    Price
  • AC Adapter (for Continuous Display)
    Discontinued
    Supplier Number: -
    Spécifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    12444-120
    W×D×H
    Small Business Enterprise
  • FLASH Memory Card
    T: –184…+1093 °C (–300…+2000 °F)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $87.23
    $87.23
    Each
       $87.23
    Supplier Number: A210
    Spécifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    14905-018
    W×D×H
    Ø 150 mm (6")
    Small Business Enterprise
  • Memory Card Reader Kit
    T: –45…+176 °C (–50…+350 °F)
    Discontinued
    Supplier Number: -
    Spécifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    14905-020
    W×D×H
    Ø 150 mm (6")
    Small Business Enterprise