Polypropylene Screw Caps for Dropper Tips, Wheaton
13–425, White
Supplier: DWK Life Sciences (KIMBLE)
Fournisseur #: W242540-A
Polypropylene Screw Caps for Dropper Tips, Wheaton
Catalog # 89048-952
Supplier: DWK Life Sciences (KIMBLE)
Jump to:
Product Details & Documents
Caps with extension to cover dropper tip screw onto the outside of the bottle neck and are available in bulk
Use to cover Wheaton dropper tips (series 89048-954).
Spécifications
- Taille du bouchon:13–425
- Color:White