NALGENE® Polypropylene Screw Closures, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
À propos de cet article
Caps for large NALGENE® bottles and carboys
Note: Before autoclaving, set cap on top of container without engaging threads.
Caps 16225-204 and -234 are buttress-style. Autoclavable.
Frequently Bought Together