VWR® Freezer Racks
VWR® Freezer Racks
VWR® Freezer Racks
VWR® Freezer Racks
Corrosion-resistant, stainless steel horizontal racks feature handles for easy transport.
  • Corrosion-resistant
  • Stainless steel fabrication
  • Multiple shelf counts
Three-shelf rack fits in upright freezers and holds one 7.6 cm (3") box per shelf. Four-shelf rack fits in chest freezers and holds one 5.1 cm (2") box per shelf.
Product Family Options
  • Désignation
    L×D×H
  • Three-Shelf Rack, 3" Boxes
    22 x 5.5 x 8.9"
    Each
    Each - 402,63 $
    402,63 $
    Each
       402,63 $
  • Four-Shelf Rack, 2" Boxes
    21.8 x 5.5 x 8.9"
    Each
    Each - 448,88 $
    448,88 $
    Each
       448,88 $