SP Bel-Art No-Wire™ Multi-Tube Racks, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
À propos de cet article
Choose from two space-saving rack configurations, as both styles accommodate round and conical bottom tubes.
- White polypropylene construction
- Autoclavable
- Easy to assemble
- Space-saving design
Made of white polypropylene, they are steam autoclavable at 121°C (250°F). Racks are shipped flat and assemble easily.
Frequently Bought Together