Evolution® Medium-Height Workstations, Kewaunee® | Avantor
Evolution® Medium-Height Workstations, Kewaunee®
Supplier:   Kewaunee
Evolution® Medium-Height Workstations, Kewaunee®
Supplier:  Kewaunee
Certificates
Evolution® Medium-Height Workstations, Kewaunee®
Evolution® Medium-Height Workstations, Kewaunee®
4 Options Available Below
Product Details & Documents
With a maximum height of 91.4 cm (36"), the starter workbenches are perfect for technical work.
  • Features a front radius edge worksurface
Starter workbenches come with two support columns, a front apron, a modesty panel, and all hardware. Add-on workstation includes all items noted above except one supporting column and base; this workstation shares the column and base from an existing starter workbench. Worksurfaces are available in black epoxy resin, ESD almond static-dissipative laminate with grounding kit, and wallaby plastic laminate.
Ordering information: Starter workbenches and add-on workstation do not include a work surface. Work surfaces are available separately. A starter workbench is required before purchasing an add-on workstation. Standard color is light neutral with a black accent strip; for other color options, contact your Avantor sales representative.
Product Family Options
Informations sur le produit
  • Désignation
    Hauteur
    W×D
    Availability
    Price
  • Starter workbench
    36"
    48×30"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,519.31
    $3,519.31
    Each
       $3,519.31
    La quantité minimale de commande est de 1
    Supplier Number: V407314NTK
  • Starter workbench
    36"
    60×30"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,786.82
    $4,786.82
    Each
       $4,786.82
    La quantité minimale de commande est de 1
    Supplier Number: V407315NTK
  • Starter workbench
    36"
    72×30"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,881.36
    $4,881.36
    Each
       $4,881.36
    La quantité minimale de commande est de 1
    Supplier Number: V407316NTK
  • Add-on workstation
    36"
    60×30"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,574.64
    $2,574.64
    Each
       $2,574.64
    La quantité minimale de commande est de 1
    Supplier Number: V407318NTK