Evolution® High-Column Workstations, Kewaunee®
Supplier:   Kewaunee
Evolution® High-Column Workstations, Kewaunee®
Supplier:  Kewaunee
Certificates
Evolution® High-Column Workstations, Kewaunee®
Evolution® High-Column Workstations, Kewaunee®
4 Options Available Below
Product Details & Documents
With a maximum height of 193 cm (76"), this workstation allows room for overhead storage, shelving, and lights.
  • The station features a front radius edge work surface plus two supporting columns with bases and glides
Starter workbenches include two columns, a modesty panel, front apron, worksurface brackets, and all hardware. Add-on workstation includes all items noted above except one supporting column and base; this workstation shares the column and base from an existing starter workbench.
Work surfaces are available in black epoxy resin, ESD almond static-dissipative laminate with grounding kit, and wallaby plastic laminate.
Depth: 30"
 : Starter workbenches and add-on workstation do not include a work surface. Work surfaces are available separately. A starter workbench is required before purchasing an add-on workstation. Standard color is light neutral with a black accent strip; or other color options, contact your Avantor sales representative.
 : Support brackets, an apron, and a modesty panel are included.
Product Family Options
Product Information
  • Désignation
    Hauteur
    W×D
    Availability
    Price
  • Workstation with gray ESD work surface
    76"
    48×30"
    Discontinued
    Supplier Number: -
  • Workstation with gray ESD work surface
    76"
    60×30"
    Discontinued
    Supplier Number: -
  • Workstation with gray ESD work surface
    76"
    72×30"
    Discontinued
    Supplier Number: -
  • Starter workbench
    76"
    48×30"
    Log in for availability
    Log in for availability
    Each
    Each - $4,755.85
    $4,755.85
    Each
       $4,755.85
    Supplier Number: V407324NTK
  • Starter workbench
    76"
    60×30"
    Log in for availability
    Log in for availability
    Each
    Each - $4,886.60
    $4,886.60
    Each
       $4,886.60
    Supplier Number: V407325NTK
  • Starter workbench
    76"
    72×30"
    Log in for availability
    Log in for availability
    Each
    Each - $7,406.62
    $7,406.62
    Each
       $7,406.62
    Supplier Number: V407326NTK
  • Add-on workstation
    76"
    48×30"
    Log in for availability
    Log in for availability
    Each
    Each - $4,171.79
    $4,171.79
    Each
       $4,171.79
    Supplier Number: V407327NTK
  • Almond ESD plastic laminate work surface, with grounding kit
    36"
    48×30"
    Discontinued
    Supplier Number: -
  • Black epoxy resin work surface
    36"
    60×30"
    Discontinued
    Supplier Number: -
  • Wallaby plastic laminate work surface
    36"
    72×30"
    Discontinued
    Supplier Number: -
  • Almond ESD plastic laminate work surface, with grounding kit
    36"
    60×30"
    Discontinued
    Supplier Number: -
  • Black epoxy resin work surface
    15"
    48×30"
    Discontinued
    Supplier Number: -
  • Wallaby plastic laminate work surface
    15"
    60×30"
    Discontinued
    Supplier Number: -
  • Almond ESD plastic laminate work surface, with grounding kit
    15"
    72×30"
    Discontinued
    Supplier Number: -
  • Black epoxy resin work surface
    33"
    48×30"
    Discontinued
    Supplier Number: -
  • Wallaby plastic laminate work surface
    30"
    48×30"
    Discontinued
    Supplier Number: -