VWR® Bunsen Burners
VWR® Bunsen Burners
VWR® Bunsen Burners
Supplier:  VWR International
Certificates
VWR® Bunsen Burners
VWR® Bunsen Burners
2 Options Available Below
Product Details & Documents
Designed with a flame stabilizer to provide a steady flame for general laboratory needs.
  • Stable, heavy base
  • Steady flame
Stable, heavyweight die-cast base of nickel-plated zinc-alloy with aluminum mixing tube, 8mm (5/16") serrated gas inlet, and air regulator. The optional wing top produces a broad flat flame for working glass and general lab use.
Ordering information: Shipped complete. The wing top must be ordered separately.
Product Family Options
Product Information
  • Désignation
    For
    Output
    Hauteur
    Availability
    Price
  • VWR® Bunsen burner
    Natural gas
    800–1200 BTU
    15.6 cm (6⅛")
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $298.57
    $298.57
    New Customer Promotional Price
    Log in for availability
    Case of 10
    Case of 10 - $2,985.68
    $2,985.68
    New Customer Promotional Price
    Each
       $298.57
    Case of 10
       $2,985.68
  • VWR® Bunsen burner
    Liquid propane
    2000–3200 BTU
    15.6 cm (6⅛")
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $282.06
    $282.06
    New Customer Promotional Price
    Log in for availability
    Case of 10
    Case of 10 - $2,820.64
    $2,820.64
    New Customer Promotional Price
    Each
       $282.06
    Case of 10
       $2,820.64