Snap-On Stoppers, WHEATON®
Supplier: DWK Life Sciences (KIMBLE)
Jump to:
À propos de cet article
These snap-on stoppers provide maximum sealing for vials with a blow-back glass finish, which is a small ridge on the interior of the vials neck.
- Not recommended for vials with a straight-walled finish
The blow-back feature is typically found in vials from North American manufacturers.
The snap-on stopper locks into the blow-back ridge, providing a tight seal and keeping the stopper from backing out.
4 Options Available Below
Product Details & Documents
These snap-on stoppers provide maximum sealing for vials with a blow-back glass finish, which is a small ridge on the interior of the vials neck.
- Not recommended for vials with a straight-walled finish
The blow-back feature is typically found in vials from North American manufacturers.
The snap-on stopper locks into the blow-back ridge, providing a tight seal and keeping the stopper from backing out.
Product Family Options
Informations sur le produit
- DésignationTailleMatièreColorAvailabilityPrice
- Snap-on stopper7×13 mmRubberRedLog in pour la disponibilitéLog in pour la disponibilitéCase of 1,000Case of 1,000 - $529.81$529.81Prix promotionnel pour les nouveaux clientsCase of 1,000$529.81Supplier Number: 224100-080
- Snap-on stopper7×3 mmButylGrayLog in pour la disponibilitéLog in pour la disponibilitéCase of 1,000Case of 1,000 - $384.20$384.20Prix promotionnel pour les nouveaux clientsCase of 1,000$384.20Supplier Number: W224100-081
- Snap-on stopper13×20 mmRubberRedLog in pour la disponibilitéLog in pour la disponibilitéCase of 1,000Case of 1,000 - $580.55$580.55Prix promotionnel pour les nouveaux clientsCase of 1,000$580.55Supplier Number: 224100-180
- Snap-on stopper13×20 mmButylGrayLog in pour la disponibilitéLog in pour la disponibilitéCase of 1,000Case of 1,000 - $570.64$570.64Prix promotionnel pour les nouveaux clientsCase of 1,000$570.64Supplier Number: W224100-181