VWR® Basic SPP Shoe Covers
VWR® Basic Protection SPP Shoe Covers are manufactured from strong, lightweight, and breathable spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates in non-hazardous environments. These shoe covers are ideal for less critical areas or pre-gowning entry rooms.
Breathable and low lint SPP material maintains cleanroom integrity while allowing airflow
- Elastic top and bottom ensures a secure fit over various shoe sizes
- Latex-free construction is safe for users with latex sensitivities
- Ideal for non-hazardous environments
- Available with non-skid sole to minimize risk of slips in cleanroom or lab settings
- Cost-effective disposable garments eliminate laundering and reduce contamination risk
- Lot-controlled and manufactured in ISO certified facilities
VWR® Basic Protection SPP Shoe Covers are engineered to provide dependable protection in cleanrooms, laboratories, and other controlled environments where contamination control is critical. Constructed from 35 gsm spunbonded polypropylene (SPP), these covers offer a lightweight yet durable barrier against dry particulates and non-hazardous materials. The breathable, low-lint fabric ensures comfort during extended wear while maintaining the integrity of sensitive environments.
For added safety, shoe covers are available with a non-skid rubber-coated sole that enhances traction and reduces slip hazards on smooth surfaces. Elastic openings at both the top and bottom provide a secure fit over a wide range of footwear sizes, ensuring convenience and reliability. Manufactured in ISO13485:2016-certified facilities and classified as FDA Class I devices, these covers meet stringent quality and compliance standards. They are also latex-free, making them suitable for users with latex sensitivities.
Available in blue or white for easy identification, VWR® Basic Protection SPP Shoe Covers deliver a cost-effective, disposable solution for maintaining cleanliness and safety without compromising comfort or performance.
: Tested and manufactured in an ISO certified facility under stringent process controls to ensure that each product meets exacting quality standards and performs to specification. Products are validated through independent lab testing.
: Shoe covers are packaged 300 per case.
VWR® Basic Protection SPP Shoe Covers are manufactured from strong, lightweight, and breathable spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates in non-hazardous environments. These shoe covers are ideal for less critical areas or pre-gowning entry rooms.
Breathable and low lint SPP material maintains cleanroom integrity while allowing airflow
- Elastic top and bottom ensures a secure fit over various shoe sizes
- Latex-free construction is safe for users with latex sensitivities
- Ideal for non-hazardous environments
- Available with non-skid sole to minimize risk of slips in cleanroom or lab settings
- Cost-effective disposable garments eliminate laundering and reduce contamination risk
- Lot-controlled and manufactured in ISO certified facilities
VWR® Basic Protection SPP Shoe Covers are engineered to provide dependable protection in cleanrooms, laboratories, and other controlled environments where contamination control is critical. Constructed from 35 gsm spunbonded polypropylene (SPP), these covers offer a lightweight yet durable barrier against dry particulates and non-hazardous materials. The breathable, low-lint fabric ensures comfort during extended wear while maintaining the integrity of sensitive environments.
For added safety, shoe covers are available with a non-skid rubber-coated sole that enhances traction and reduces slip hazards on smooth surfaces. Elastic openings at both the top and bottom provide a secure fit over a wide range of footwear sizes, ensuring convenience and reliability. Manufactured in ISO13485:2016-certified facilities and classified as FDA Class I devices, these covers meet stringent quality and compliance standards. They are also latex-free, making them suitable for users with latex sensitivities.
Available in blue or white for easy identification, VWR® Basic Protection SPP Shoe Covers deliver a cost-effective, disposable solution for maintaining cleanliness and safety without compromising comfort or performance.
: Tested and manufactured in an ISO certified facility under stringent process controls to ensure that each product meets exacting quality standards and performs to specification. Products are validated through independent lab testing.
: Shoe covers are packaged 300 per case.
- Catalog No:
- 97041-234
- 414004-653
- 97041-236
- 89233-794
- 89233-796
- Fabric:
- Spunbound polypropylene
- Spunbound polypropylene
- Spunbound polypropylene
- Spunbound polypropylene
- Spunbound polypropylene
- Cleanroom class:
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- 10000/ISO 7
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Garment application:
- Cleanroom|Industrial|Laboratory|Medical|General Purpose
- Cleanroom|Industrial|Laboratory|Medical|General Purpose
- Cleanroom|Industrial|Laboratory|Medical|General Purpose
- Cleanroom|Industrial|Laboratory|Medical|General Purpose
- Cleanroom|Industrial|Laboratory|Medical|General Purpose
- Bacterial filtration efficiency:
- 0.55000000000000004
- 0.55000000000000004
- 0.55000000000000004
- 0.55000000000000004
- 0.55000000000000004
- Particle shedding (Helmke Drum):
- Level II
- Level II
- Level II
- Level II
- Level II
- Water resistance:
- First line barrier
- First line barrier
- First line barrier
- —
- —
- Tearing strength:
- 6.1 lbs./in. (lengthwise yarns), 7.9 lbs./in. (widthwise yarns)
- 6.1 lbs./in. (lengthwise yarns), 7.9 lbs./in. (widthwise yarns)
- 6.1 lbs./in. (lengthwise yarns), 7.9 lbs./in. (widthwise yarns)
- —
- —
- Weight:
- 36.89 g/m²
- 36.89 g/m²
- 36.89 g/m²
- —
- —
Product Family Options
Standard shoe covers
- DésignationColorPointureHauteurLongueurØ
- Standard shoe coversBlueUniversal6¾"17½"5" (top relaxed), 11½" (top stretched)Case of 300$47.34Pallet of 16200$2,556.58
Specifications
Cat. No.97041-234FabricSpunbound polypropyleneCleanroom class10000/ISO 7GenderUnisexGarment applicationCleanroom|Industrial|Laboratory|Medical|General PurposeBacterial filtration efficiency0.55000000000000004Particle shedding (Helmke Drum)Level IIWater resistanceFirst line barrierTearing strength6.1 lbs./in. (lengthwise yarns), 7.9 lbs./in. (widthwise yarns)Weight36.89 g/m²
Anti-skid shoe covers
- DésignationColorPointureHauteurLongueurØ
- Anti-skid shoe coversBlueXL7¾"18½"5½" (top relaxed), 15" (top stretched)Case of 300$174.87Pallet of 14400$8,393.86
Specifications
Cat. No.414004-653FabricSpunbound polypropyleneCleanroom class10000/ISO 7GenderUnisexGarment applicationCleanroom|Industrial|Laboratory|Medical|General PurposeBacterial filtration efficiency0.55000000000000004Particle shedding (Helmke Drum)Level IIWater resistanceFirst line barrierTearing strength6.1 lbs./in. (lengthwise yarns), 7.9 lbs./in. (widthwise yarns)Weight36.89 g/m² - Anti-skid shoe coversBlueUniversal6¾"17½"5" (top relaxed), 11½" (top stretched)Case of 300$131.24Pallet of 18000$7,874.40
Specifications
Cat. No.97041-236FabricSpunbound polypropyleneCleanroom class10000/ISO 7GenderUnisexGarment applicationCleanroom|Industrial|Laboratory|Medical|General PurposeBacterial filtration efficiency0.55000000000000004Particle shedding (Helmke Drum)Level IIWater resistanceFirst line barrierTearing strength6.1 lbs./in. (lengthwise yarns), 7.9 lbs./in. (widthwise yarns)Weight36.89 g/m² - Heavy weight anti-skid shoe coversBlueUniversal—15½"—-
Specifications
Cat. No.89233-794FabricSpunbound polypropyleneCleanroom class10000/ISO 7GenderUnisexGarment applicationCleanroom|Industrial|Laboratory|Medical|General PurposeBacterial filtration efficiency0.55000000000000004Particle shedding (Helmke Drum)Level IIWater resistance—Tearing strength—Weight— - Heavy weight anti-skid shoe coversBlueXL—17"—-
Specifications
Cat. No.89233-796FabricSpunbound polypropyleneCleanroom class10000/ISO 7GenderUnisexGarment applicationCleanroom|Industrial|Laboratory|Medical|General PurposeBacterial filtration efficiency0.55000000000000004Particle shedding (Helmke Drum)Level IIWater resistance—Tearing strength—Weight—