Stain Kits, BD Diagnostics
Stain Kits, BD Diagnostics
Supplier:   BD
Stain Kits, BD Diagnostics
Supplier:  BD
Certificates
Stain Kits, BD Diagnostics
3 Options Available Below
Product Details & Documents
These kits include all the necessary components for categorizing bacteria as gram-negative or gram-positive, a definitive characteristic in the organism identification process.
Fluorescent stain kit 90008-886 enables the observation of smears at a lower magnification, increasing the field of view and decreasing examination time. Kit includes one 250mL bottle each of the following: TB Auramine-Rhodamine T, TB Decolorizer TM, and TB Potassium Permanganate.
TB stain kit K 90008-888 includes one 250mL bottle each of TB Carbolfuschin KF, TB Decolorizer, and TB Brilliant Green K.
TB stain kit ZN 90008-884 includes one 250mL bottle each of TB Carbolfuschin ZN, TB Decolorizer, and TB Methylene Blue.
Ordering information: All stain kits come conveniently packaged in a compact tray and bottles feature no-drip “keep clean” cap spouts for dispensing.
Product Family Options
Product Information
  • Désignation
    Taille
    Availability
    Price
  • TB Stain Kit ZN
    250 ml (8.4 oz.)
    Log in for availability
    Log in for availability
    Each
    Each - $262.00
    $262.00
    Each
       $262.00
    Supplier Number: 212520
  • TB Fluorescent Stain Kit T
    250 ml (8.4 oz.)
    Log in for availability
    Log in for availability
    Each (250ml)
    Each (250ml) - $331.00
    $331.00
    Each (250ml)
       $331.00
    Supplier Number: 212521
  • TB Stain Kit K
    250 ml (8.4 oz.)
    Log in for availability
    Log in for availability
    Each
    Each - $273.00
    $273.00
    Each
       $273.00
    Supplier Number: 212522