InterPed™ Impervious Shoe Covers, InterWorld™
InterPed™ Impervious Shoe Covers, InterWorld™
Supplier:   Interworld
InterPed™ Impervious Shoe Covers, InterWorld™
Supplier:  Interworld
Certificates
InterPed™ Impervious Shoe Covers, InterWorld™
InterPed™ Impervious Shoe Covers, InterWorld™
2 Options Available Below
Product Details & Documents
Heavyweight shoe covers effectively contain dirt and moisture
They are 40.6cm (16") long and made of compressed polyethylene. Shoe covers feature seamless bottoms and are nontoxic and odorless.
Ordering information: Packed 100 shoe covers per bag, 3 bags per case.
Specifications for Product Family
Specifications
  • Catalog No:
  • 21923-234
  • 21923-238
  • Fabric:
  • Polyethylene
  • Polyethylene
  • Garment application:
  • Laboratory
  • Laboratory
Specifications
Product Family Options
Product Information
  • Color
    Longueur
    Availability
    Price
  • White
    40.6 cm (16")
    Log in for availability
    Log in for availability
    Case of 300
    Case of 300 - $1,128.30
    $1,128.30
    Case of 300
       $1,128.30
    Supplier Number: 10135-107-02
    Specifications:
    More
    Specifications
    Cat. No.
    21923-234
    Fabric
    Polyethylene
    Garment application
    Laboratory
  • Blue
    40.6 cm (16")
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $351.15
    $351.15
    Case of 1,000
       $351.15
    Supplier Number: 10135-136-02
    Specifications:
    More
    Specifications
    Cat. No.
    21923-238
    Fabric
    Polyethylene
    Garment application
    Laboratory