InterPed™ TRIBO™ Shoe Covers, InterWorld™
White, 40.6 cm (16")
Catalog # 21923-274
Supplier: Interworld
Fournisseur #: 10070-387-02
Jump to:
Product Details & Documents
TRIBO™ nonskid shoe covers offer excellent traction on vinyl floors and most other walking surfaces
Fluid-resistant shoe covers are constructed of nonwoven spunbonded polypropylene coated with a proprietary elastomer blend that is impervious to water, grease, and oil.
Ordering information: Packaged in a cleanroom environment, 100 shoe covers per bag, 3 bags per case.
Spécifications
- Color:White
- Longueur:40.6 cm (16")
- Cat. No.:21923-274
- Garment application:Laboratory