VWR® Basic SPP Beard Covers
VWR® Basic SPP Beard Covers
VWR® Basic SPP Beard Covers
Supplier:  VWR International
Certificates
VWR® Basic SPP Beard Covers
2 Options Available Below
Product Details & Documents
VWR® Basic protection SPP beard covers are manufactured from a spunbonded polypropylene (SPP) fabric that is strong, lightweight, and breathable
  • Economical first-line barrier
  • Non-linting
Elastic band provides secure fit. Available in latex or latex-free formulation. Beard covers are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validated through independent lab testing.
They protect against dirt, grime, and certain dry particulates in nonhazardous environments. These general-purpose garments are ideal for less critical areas or pre-gowning entry rooms.
Documents
Specifications for Product Family
Specifications
  • Catalog No:
  • 89107-778
  • 89218-950
  • 89107-776
  • 76019-100
  • Particle shedding (Helmke Drum):
  • Level II
  • Level II
  • Level II
  • Tensile strength:
  • >30 (Elongation Percentage)
  • >30 (Elongation Percentage)
  • >30 (Elongation Percentage)
  • Weight:
  • 10.0 g/m²
  • 10.0 g/m2
  • 10.0 g/m²
Specifications
Product Family Options
Product Information
  • Désignation
    Color
    Taille de confection
    Longueur
    Availability
    Price
  • Latex-free
    White
    Taille unique
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $179.74
    $179.74
    New Customer Promotional Price
    Log in for availability
    Pallet of 64000
    Pallet of 64000 - $11,503.10
    $11,503.10
    New Customer Promotional Price
    Case of 1,000
       $179.74
    Pallet of 64000
       $11,503.10
    Specifications:
    More
    Specifications
    Cat. No.
    89107-778
    Particle shedding (Helmke Drum)
    Level II
    Tensile strength
    >30 (Elongation Percentage)
    Weight
    10.0 g/m²
  • Latex-free
    White
    XL
    18"
    Log in for availability
    Log in for availability
    Log in for availability
    Case of 1,000
    Case of 1,000 - $201.10
    $201.10
    New Customer Promotional Price
    Log in for availability
    Pallet of 64000
    Pallet of 64000 - $12,870.66
    $12,870.66
    New Customer Promotional Price
    Case of 1,000
       $201.10
    Pallet of 64000
       $12,870.66
    Specifications:
    More
    Specifications
    Cat. No.
    89218-950
    Particle shedding (Helmke Drum)
    Level II
    Tensile strength
    >30 (Elongation Percentage)
    Weight
    10.0 g/m2
  • With Latex
    White
    Taille unique
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    89107-776
    Particle shedding (Helmke Drum)
    Level II
    Tensile strength
    >30 (Elongation Percentage)
    Weight
    10.0 g/m²
  • Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    76019-100
    Particle shedding (Helmke Drum)
    Tensile strength
    Weight