VWR® Basic Protection SMS Lab Coats
VWR® Basic Protection SMS Lab Coats
New Product
Supplier:   VWR International
VWR® Basic Protection SMS Lab Coats
Supplier:  VWR International
New Product
Certificates
VWR® Basic Protection SMS Lab Coats
17 Options Available Below
Product Details & Documents
VWR® Basic protection SMS lab coats are designed to provide lightweight, breathable coverage for non-hazardous environments. Constructed from SMS (Spunbond-Meltblown-Spunbond) material, they offer a reliable barrier against dust, dirt, and light liquid splashes while maintaining comfort during extended wear.
  • Lightweight, breathable, and durable SMS material for comfort during extended wear
  • Snap-front closure is easy to put on and remove while maintaining a secure fit
  • Collar design provides added coverage and a professional appearance
  • Shields against dust, dirt, and light liquid splashes in non-hazardous environments
  • Breathable fabric reduces heat buildup and improves wearer comfort over extended periods
  • Ideal for routine lab work and controlled environments without over-specifying protection
VWR® Basic protection SMS lab coats are engineered for environments where comfort and essential coverage are key. Constructed from SMS (Spunbond-Meltblown-Spunbond) fabric, these coats combine three layers of polypropylene to deliver a balance of strength, breathability, and barrier protection. The outer spunbond layers provide durability and resistance to tearing, while the meltblown middle layer acts as a filter against dust, dirt, and light liquid splashes. This makes them ideal for non-hazardous applications such as general laboratory work, cleanroom support tasks, and production areas where particulate control is important but chemical exposure is minimal.
Designed with practical features, these lab coats include a secure snap-front closure for easy donning and removal, a collar for added coverage, and elastic cuffs to ensure a snug fit that helps maintain cleanliness. Their lightweight construction promotes comfort during extended wear, reducing heat buildup and improving mobility. Available in multiple sizes, they support compliance with workplace safety protocols while maintaining a professional appearance. For facilities seeking cost-effective, disposable apparel that meets basic protection needs without sacrificing comfort, VWR® SMS lab coats offer a dependable solution.
Packaging: Lab coats are packaged 30 per case.
Documents
Specifications for Product Family
Spécifications
  • Catalog No:
  • 414004-359
  • 78021-804
  • 414004-366
  • 414004-358
  • 414004-362
  • 414004-355
  • 414004-363
  • 414004-356
  • 414004-361
  • 414004-357
  • 414004-364
  • 414004-360
  • 414004-365
  • 414004-367
  • 75840-186
  • 75854-026
  • 75854-028
  • Fabric:
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Spunbound polypropylene
  • Closure type:
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Cleanroom class:
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • Pockets:
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Sleeve or leg length:
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Garment application:
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Air permeability:
  • 95.3 cu. ft./min./sq. ft.
  • 95.3 cu. ft./min./sq. ft.
  • 95.3 cu. ft./min./sq. ft.
  • 95.3 cu. ft./min./sq. ft.
  • 95.3 cu. ft./min./sq. ft.
  • 95.3 cu. ft./min./sq. ft.
  • 95.3 cu. ft./min./sq. ft.
  • 95.3 cu. ft./min./sq. ft.
  • 95.3 cu. ft./min./sq. ft.
  • 95.3 cu. ft./min./sq. ft.
  • 95.3 cu. ft./min./sq. ft.
  • 95.3 cu. ft./min./sq. ft.
  • 95.3 cu. ft./min./sq. ft.
  • 95.3 cu. ft./min./sq. ft.
  • 95.3 cu. ft./min./sq. ft.
  • 95.3 cu. ft./min./sq. ft.
  • 95.3 cu. ft./min./sq. ft.
  • Bacterial filtration efficiency:
  • 90%
  • 90%
  • 90%
  • 90%
  • 90%
  • 90%
  • 90%
  • 90%
  • 90%
  • 90%
  • 90%
  • 90%
  • 90%
  • 90%
  • 90%
  • 90%
  • 90%
  • Particle shedding (Helmke Drum):
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Water resistance:
  • 16.7 G
  • 16.7 G
  • 16.7 G
  • 16.7 G
  • 16.7 G
  • 16.7 G
  • 16.7 G
  • 16.7 G
  • 16.7 G
  • 16.7 G
  • 16.7 G
  • 16.7 G
  • 16.7 G
  • 16.7 G
  • 16.7 G
  • 16.7 G
  • 16.7 G
  • Water vapor transmission rate:
  • 965 g/m²/24 h
  • 965 g/m²/24 h
  • 965 g/m²/24 h
  • 965 g/m²/24 h
  • 965 g/m²/24 h
  • 965 g/m²/24 h
  • 965 g/m²/24 h
  • 965 g/m²/24 h
  • 965 g/m²/24 h
  • 965 g/m²/24 h
  • 965 g/m²/24 h
  • 965 g/m²/24 h
  • 965 g/m²/24 h
  • 965 g/m²/24 h
  • 965 g/m²/24 h
  • 965 g/m²/24 h
  • 965 g/m²/24 h
  • Tensile strength:
  • 13.0 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13.0 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13.0 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13.0 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13.0 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13.0 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13.0 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13.0 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13.0 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13.0 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13.0 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13.0 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13.0 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13.0 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13.0 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13.0 lbs./in. (warp), 16.1 lbs./in. (filling)
  • 13.0 lbs./in. (warp), 16.1 lbs./in. (filling)
  • Tearing strength:
  • 3.1 lbs./in. (lengthwise yarns), 2.6 lbs./in. (widthwise yarns)
  • 3.1 lbs./in. (lengthwise yarns), 2.6 lbs./in. (widthwise yarns)
  • 3.1 lbs./in. (lengthwise yarns), 2.6 lbs./in. (widthwise yarns)
  • 3.1 lbs./in. (lengthwise yarns), 2.6 lbs./in. (widthwise yarns)
  • 3.1 lbs./in. (lengthwise yarns), 2.6 lbs./in. (widthwise yarns)
  • 3.1 lbs./in. (lengthwise yarns), 2.6 lbs./in. (widthwise yarns)
  • 3.1 lbs./in. (lengthwise yarns), 2.6 lbs./in. (widthwise yarns)
  • 3.1 lbs./in. (lengthwise yarns), 2.6 lbs./in. (widthwise yarns)
  • 3.1 lbs./in. (lengthwise yarns), 2.6 lbs./in. (widthwise yarns)
  • 3.1 lbs./in. (lengthwise yarns), 2.6 lbs./in. (widthwise yarns)
  • 3.1 lbs./in. (lengthwise yarns), 2.6 lbs./in. (widthwise yarns)
  • 3.1 lbs./in. (lengthwise yarns), 2.6 lbs./in. (widthwise yarns)
  • 3.1 lbs./in. (lengthwise yarns), 2.6 lbs./in. (widthwise yarns)
  • 3.1 lbs./in. (lengthwise yarns), 2.6 lbs./in. (widthwise yarns)
  • 3.1 lbs./in. (lengthwise yarns), 2.6 lbs./in. (widthwise yarns)
  • 3.1 lbs./in. (lengthwise yarns), 2.6 lbs./in. (widthwise yarns)
  • 3.1 lbs./in. (lengthwise yarns), 2.6 lbs./in. (widthwise yarns)
  • Weight:
  • 50.45 g/m²
  • 50.45 g/m²
  • 50.45 g/m²
  • 50.45 g/m²
  • 50.45 g/m²
  • 50.45 g/m²
  • 50.45 g/m²
  • 50.45 g/m²
  • 50.45 g/m²
  • 50.45 g/m²
  • 50.45 g/m²
  • 50.45 g/m²
  • 50.45 g/m²
  • 50.45 g/m²
  • 50.45 g/m²
  • 50.45 g/m²
  • 50.45 g/m²
Spécifications
Product Family Options
Light blue
Informations sur le produit
  • Désignation
    Color
    Taille de confection
    Longueur
    Chest Size
    Sleeve
    Availability
    Price
  • Bleu clair
    S
    36¾"
    24¾"
    30"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $363.94
    $363.94
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 720
    Pallet of 720 - $8,734.66
    $8,734.66
    Prix promotionnel pour les nouveaux clients
    Case of 30
       $363.94
    Pallet of 720
       $8,734.66
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-359
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Air permeability
    95.3 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    90%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    16.7 G
    Water vapor transmission rate
    965 g/m²/24 h
    Tensile strength
    13.0 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise yarns), 2.6 lbs./in. (widthwise yarns)
    Weight
    50.45 g/m²
  • Bleu clair
    M
    39½"
    25¼"
    30¼"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $365.28
    $365.28
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 720
    Pallet of 720 - $8,766.72
    $8,766.72
    Prix promotionnel pour les nouveaux clients
    Case of 30
       $365.28
    Pallet of 720
       $8,766.72
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-358
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Air permeability
    95.3 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    90%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    16.7 G
    Water vapor transmission rate
    965 g/m²/24 h
    Tensile strength
    13.0 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise yarns), 2.6 lbs./in. (widthwise yarns)
    Weight
    50.45 g/m²
  • Bleu clair
    L
    40"
    27"
    32½"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $368.79
    $368.79
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 750
    Pallet of 750 - $9,219.80
    $9,219.80
    Prix promotionnel pour les nouveaux clients
    Case of 30
       $368.79
    Pallet of 750
       $9,219.80
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-355
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Air permeability
    95.3 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    90%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    16.7 G
    Water vapor transmission rate
    965 g/m²/24 h
    Tensile strength
    13.0 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise yarns), 2.6 lbs./in. (widthwise yarns)
    Weight
    50.45 g/m²
  • Bleu clair
    XL
    42¾"
    27¾"
    33"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $359.45
    $359.45
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 720
    Pallet of 720 - $8,626.75
    $8,626.75
    Prix promotionnel pour les nouveaux clients
    Case of 30
       $359.45
    Pallet of 720
       $8,626.75
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-356
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Air permeability
    95.3 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    90%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    16.7 G
    Water vapor transmission rate
    965 g/m²/24 h
    Tensile strength
    13.0 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise yarns), 2.6 lbs./in. (widthwise yarns)
    Weight
    50.45 g/m²
  • Bleu clair
    XXL
    43½"
    29½"
    37½"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $371.74
    $371.74
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 720
    Pallet of 720 - $8,921.66
    $8,921.66
    Prix promotionnel pour les nouveaux clients
    Case of 30
       $371.74
    Pallet of 720
       $8,921.66
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-357
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Air permeability
    95.3 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    90%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    16.7 G
    Water vapor transmission rate
    965 g/m²/24 h
    Tensile strength
    13.0 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise yarns), 2.6 lbs./in. (widthwise yarns)
    Weight
    50.45 g/m²
  • Bleu clair
    3XL
    44½"
    30¾"
    38¼"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $376.23
    $376.23
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 720
    Pallet of 720 - $9,029.57
    $9,029.57
    Prix promotionnel pour les nouveaux clients
    Case of 30
       $376.23
    Pallet of 720
       $9,029.57
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-360
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Air permeability
    95.3 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    90%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    16.7 G
    Water vapor transmission rate
    965 g/m²/24 h
    Tensile strength
    13.0 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise yarns), 2.6 lbs./in. (widthwise yarns)
    Weight
    50.45 g/m²
  • Light blue
    4XL
    46½"
    31½"
    39¼"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $371.10
    $371.10
    Prix promotionnel pour les nouveaux clients
    Case of 30
       $371.10
    Spécifications:
    More
    Specifications
    Cat. No.
    75840-186
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Air permeability
    95.3 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    90%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    16.7 G
    Water vapor transmission rate
    965 g/m²/24 h
    Tensile strength
    13.0 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise yarns), 2.6 lbs./in. (widthwise yarns)
    Weight
    50.45 g/m²
  • Light blue
    XS
    35½"
    24"
    29¼"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $365.14
    $365.14
    Prix promotionnel pour les nouveaux clients
    Case of 30
       $365.14
    Spécifications:
    More
    Specifications
    Cat. No.
    75854-026
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Air permeability
    95.3 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    90%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    16.7 G
    Water vapor transmission rate
    965 g/m²/24 h
    Tensile strength
    13.0 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise yarns), 2.6 lbs./in. (widthwise yarns)
    Weight
    50.45 g/m²
  • Light blue
    5XL
    47¾"
    32¼"
    40¼"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $382.76
    $382.76
    Prix promotionnel pour les nouveaux clients
    Case of 30
       $382.76
    Spécifications:
    More
    Specifications
    Cat. No.
    75854-028
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Air permeability
    95.3 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    90%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    16.7 G
    Water vapor transmission rate
    965 g/m²/24 h
    Tensile strength
    13.0 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise yarns), 2.6 lbs./in. (widthwise yarns)
    Weight
    50.45 g/m²
White
Informations sur le produit
  • Désignation
    Color
    Taille de confection
    Longueur
    Chest Size
    Sleeve
    Availability
    Price
  • White
    5XL
    47¾"
    32¼"
    40¼"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $228.71
    $228.71
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 750
    Pallet of 750 - $6,861.36
    $6,861.36
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $228.71
    Pallet of 750
       $6,861.36
    Spécifications:
    New Product
    More
    Specifications
    Cat. No.
    78021-804
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Air permeability
    95.3 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    90%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    16.7 G
    Water vapor transmission rate
    965 g/m²/24 h
    Tensile strength
    13.0 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise yarns), 2.6 lbs./in. (widthwise yarns)
    Weight
    50.45 g/m²
    New Product
  • White
    S
    36¾"
    24¾"
    30"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $362.29
    $362.29
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 720
    Pallet of 720 - $8,694.91
    $8,694.91
    Prix promotionnel pour les nouveaux clients
    Case of 30
       $362.29
    Pallet of 720
       $8,694.91
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-366
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Air permeability
    95.3 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    90%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    16.7 G
    Water vapor transmission rate
    965 g/m²/24 h
    Tensile strength
    13.0 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise yarns), 2.6 lbs./in. (widthwise yarns)
    Weight
    50.45 g/m²
  • White
    M
    39½"
    25¼"
    30¼"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $365.88
    $365.88
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 720
    Pallet of 720 - $8,781.12
    $8,781.12
    Prix promotionnel pour les nouveaux clients
    Case of 30
       $365.88
    Pallet of 720
       $8,781.12
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-362
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Air permeability
    95.3 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    90%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    16.7 G
    Water vapor transmission rate
    965 g/m²/24 h
    Tensile strength
    13.0 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise yarns), 2.6 lbs./in. (widthwise yarns)
    Weight
    50.45 g/m²
  • White
    L
    40"
    27"
    32½"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $365.86
    $365.86
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 720
    Pallet of 720 - $8,780.54
    $8,780.54
    Prix promotionnel pour les nouveaux clients
    Case of 30
       $365.86
    Pallet of 720
       $8,780.54
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-363
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Air permeability
    95.3 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    90%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    16.7 G
    Water vapor transmission rate
    965 g/m²/24 h
    Tensile strength
    13.0 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise yarns), 2.6 lbs./in. (widthwise yarns)
    Weight
    50.45 g/m²
  • White
    XL
    42¾"
    27¾"
    33"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $353.57
    $353.57
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 720
    Pallet of 720 - $8,485.63
    $8,485.63
    Prix promotionnel pour les nouveaux clients
    Case of 30
       $353.57
    Pallet of 720
       $8,485.63
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-361
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Air permeability
    95.3 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    90%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    16.7 G
    Water vapor transmission rate
    965 g/m²/24 h
    Tensile strength
    13.0 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise yarns), 2.6 lbs./in. (widthwise yarns)
    Weight
    50.45 g/m²
  • White
    XXL
    43½"
    29½"
    37½"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $369.03
    $369.03
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 720
    Pallet of 720 - $8,856.77
    $8,856.77
    Prix promotionnel pour les nouveaux clients
    Case of 30
       $369.03
    Pallet of 720
       $8,856.77
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-364
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Air permeability
    95.3 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    90%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    16.7 G
    Water vapor transmission rate
    965 g/m²/24 h
    Tensile strength
    13.0 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise yarns), 2.6 lbs./in. (widthwise yarns)
    Weight
    50.45 g/m²
  • White
    3XL
    44½"
    30¾"
    38¼"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $371.41
    $371.41
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 720
    Pallet of 720 - $8,913.79
    $8,913.79
    Prix promotionnel pour les nouveaux clients
    Case of 30
       $371.41
    Pallet of 720
       $8,913.79
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-365
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Air permeability
    95.3 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    90%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    16.7 G
    Water vapor transmission rate
    965 g/m²/24 h
    Tensile strength
    13.0 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise yarns), 2.6 lbs./in. (widthwise yarns)
    Weight
    50.45 g/m²
  • White
    4XL
    46½"
    31½"
    39¼"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $377.12
    $377.12
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 720
    Pallet of 720 - $9,050.88
    $9,050.88
    Prix promotionnel pour les nouveaux clients
    Case of 30
       $377.12
    Pallet of 720
       $9,050.88
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-367
    Fabric
    Spunbound polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Air permeability
    95.3 cu. ft./min./sq. ft.
    Bacterial filtration efficiency
    90%
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    16.7 G
    Water vapor transmission rate
    965 g/m²/24 h
    Tensile strength
    13.0 lbs./in. (warp), 16.1 lbs./in. (filling)
    Tearing strength
    3.1 lbs./in. (lengthwise yarns), 2.6 lbs./in. (widthwise yarns)
    Weight
    50.45 g/m²