VWR® Basic SPP Lab Coats
VWR® Basic SPP Lab Coats
VWR® Basic SPP Lab Coats
Supplier:  VWR International
Certificates
VWR® Basic SPP Lab Coats
24 Options Available Below
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
Disposable lab coats made from spunbond polypropylene (SPP) fabric, offering lightweight and breathable protection for non-hazardous environments. Designed for comfort and contamination control, these lab coats are ideal for clean areas, food processing, and general industrial use.
  • Spunbond Polypropylene (SPP) material is lightweight and breathable
  • Breathable fabric reduces heat buildup during extended use
  • Snap front closure for easy donning and removal
  • Available with elastic or knit cuff styles
  • Non-linting fabric helps maintain cleanliness in controlled environments
  • Single-use, disposable design for contamination control
  • Economical solution for basic protection needs
  • Disposable design simplifies protocols and reduces laundering costsSpunbond Polypropylene (SPP) material is lightweight and breathable
  • Breathable fabric reduces heat buildup during extended use
  • Snap front closure for easy donning and removal
  • Available with elastic or knit cuff styles
  • Non-linting fabric helps maintain cleanliness in controlled environments
  • Single-use, disposable design for contamination control
  • Economical solution for basic protection needs
  • Disposable design simplifies protocols and reduces laundering costs
The VWR® Basic Protection Lab Coat is designed for environments where contamination control and user comfort are essential, but exposure risks remain minimal. Constructed from spunbond polypropylene (SPP), this nonwoven fabric offers a lightweight, breathable solution that promotes airflow while maintaining cleanliness. The non-linting properties help reduce particle contamination, making it suitable for clean areas, food processing, and general industrial applications.
The lab coat features a snap-front closure for quick and secure fastening, along with elastic or knit cuffs to accommodate different glove types and ensure a comfortable fit. The disposable design supports hygiene compliance by reducing cross-contamination risks and simplifying inventory management. While not intended for hazardous chemical or liquid protection, this garment provides an effective barrier against dust, dirt, and dry particulates, making it ideal for routine tasks in controlled environments.
By combining economical single-use convenience, breathability, and basic contamination control, VWR® Basic Protection SPP Lab Coats deliver a practical solution for facilities seeking dependable protective apparel for everyday operations.
Documents
Specifications for Product Family
Spécifications
  • Catalog No:
  • 89208-750
  • 89208-752
  • 89208-754
  • 89208-756
  • 89208-812
  • 89208-814
  • 89208-816
  • 89208-818
  • 89208-820
  • 89136-824
  • 89136-828
  • 414004-345
  • 414004-344
  • 414004-342
  • 414004-343
  • 414004-348
  • 414004-346
  • 414004-353
  • 414004-351
  • 414004-350
  • 414004-349
  • 414004-352
  • 76311-258
  • 414004-354
  • Fabric:
  • Spunbond polypropylene
  • Spunbond polypropylene
  • Spunbond polypropylene
  • Spunbond polypropylene
  • Spunbond polypropylene
  • Spunbond polypropylene
  • Spunbond polypropylene
  • Spunbond polypropylene
  • Spunbond polypropylene
  • Spunbond polypropylene
  • Spunbond polypropylene
  • Spunbond polypropylene
  • Spunbond polypropylene
  • Spunbond polypropylene
  • Spunbond polypropylene
  • Spunbond polypropylene
  • Spunbond polypropylene
  • Spunbond polypropylene
  • Spunbond polypropylene
  • Spunbond polypropylene
  • Spunbond polypropylene
  • Spunbond polypropylene
  • Spunbond polypropylene
  • Spunbond polypropylene
  • Closure type:
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Cleanroom class:
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • Pockets:
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • Without pockets
  • Without pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Sleeve or leg length:
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Garment application:
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Cleanroom|Industrial|Laboratory
  • Particle shedding (Helmke Drum):
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Level II
  • Water resistance:
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • First line barrier
  • Weight:
  • 30 g/m²
  • 30 g/m²
  • 30 g/m²
  • 30 g/m²
  • 40 g/m²
  • 40 g/m²
  • 40 g/m²
  • 40 g/m²
  • 40 g/m²
  • 40 g/m²
  • 40 g/m²
  • 48 g/m²
  • 48 g/m²
  • 30 g/m²
  • 48 g/m²
  • 48 g/m²
  • 48 g/m²
  • 48 g/m²
  • 48 g/m²
  • 48 g/m²
  • 48 g/m²
  • 48 g/m²
  • 48 g/m²
  • 48 g/m²
Spécifications
Product Family Options
30 g SPP, three pockets with elastic cuffs
Informations sur le produit
  • Color
    Taille de confection
    Longueur
    Chest Size
    Sleeve
    Availability
    Price
  • Blue
    L
    40"
    31½"
    33"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $101.89
    $101.89
    Prix promotionnel pour les nouveaux clients
    Case of 30
       $101.89
    Spécifications:
    More
    Specifications
    Cat. No.
    89208-750
    Fabric
    Spunbond polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    30 g/m²
  • Blue
    XL
    42¾"
    27¾"
    33"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $103.74
    $103.74
    Prix promotionnel pour les nouveaux clients
    Case of 30
       $103.74
    Spécifications:
    More
    Specifications
    Cat. No.
    89208-752
    Fabric
    Spunbond polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    30 g/m²
  • Blue
    XXL
    43½"
    31½"
    37½"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $109.49
    $109.49
    Prix promotionnel pour les nouveaux clients
    Case of 30
       $109.49
    Spécifications:
    More
    Specifications
    Cat. No.
    89208-754
    Fabric
    Spunbond polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    30 g/m²
  • Blue
    4XL
    46 B½"
    31½"
    39¼"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $113.06
    $113.06
    Prix promotionnel pour les nouveaux clients
    Case of 30
       $113.06
    Spécifications:
    More
    Specifications
    Cat. No.
    89208-756
    Fabric
    Spunbond polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    30 g/m²
40 g SPP, three pockets with elastic cuffs
Informations sur le produit
  • Color
    Taille de confection
    Longueur
    Chest Size
    Sleeve
    Availability
    Price
  • White
    M
    39½"
    25¼"
    30¼"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $255.41
    $255.41
    Prix promotionnel pour les nouveaux clients
    Case of 30
       $255.41
    Spécifications:
    More
    Specifications
    Cat. No.
    89208-812
    Fabric
    Spunbond polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    40 g/m²
  • White
    L
    40"
    31½"
    32½"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $256.73
    $256.73
    Prix promotionnel pour les nouveaux clients
    Case of 30
       $256.73
    Spécifications:
    More
    Specifications
    Cat. No.
    89208-814
    Fabric
    Spunbond polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    40 g/m²
  • White
    XL
    42¾"
    27¾"
    33"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $257.52
    $257.52
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 900
    Pallet of 900 - $7,725.60
    $7,725.60
    Prix promotionnel pour les nouveaux clients
    Case of 30
       $257.52
    Pallet of 900
       $7,725.60
    Spécifications:
    More
    Specifications
    Cat. No.
    89208-816
    Fabric
    Spunbond polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    40 g/m²
  • White
    XXL
    43½"
    31½"
    37½"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $33.79
    $33.79
    Case of 30
       $33.79
    Spécifications:
    More
    Specifications
    Cat. No.
    89208-818
    Fabric
    Spunbond polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    40 g/m²
  • White
    4XL
    44½"
    25¼"
    38¼"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $257.26
    $257.26
    Prix promotionnel pour les nouveaux clients
    Case of 30
       $257.26
    Spécifications:
    More
    Specifications
    Cat. No.
    89208-820
    Fabric
    Spunbond polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    40 g/m²
40 g SPP, no pocket with knit cuffs
Informations sur le produit
  • Color
    Taille de confection
    Longueur
    Chest Size
    Sleeve
    Availability
    Price
  • White
    2XL
    43½"
    29½"
    37½"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $294.69
    $294.69
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 720
    Pallet of 720 - $7,072.51
    $7,072.51
    Prix promotionnel pour les nouveaux clients
    Case of 30
       $294.69
    Pallet of 720
       $7,072.51
    Spécifications:
    More
    Specifications
    Cat. No.
    89136-824
    Fabric
    Spunbond polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    40 g/m²
  • White
    5XL
    48"
    32¼"
    40½"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $297.02
    $297.02
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 720
    Pallet of 720 - $7,128.58
    $7,128.58
    Prix promotionnel pour les nouveaux clients
    Case of 30
       $297.02
    Pallet of 720
       $7,128.58
    Spécifications:
    More
    Specifications
    Cat. No.
    89136-828
    Fabric
    Spunbond polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    Without pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    40 g/m²
48 g SPP, three pockets with knit cuffs
Informations sur le produit
  • Color
    Taille de confection
    Longueur
    Chest Size
    Sleeve
    Availability
    Price
  • White
    M
    36¾"
    24¼"
    30"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $223.29
    $223.29
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 720
    Pallet of 720 - $5,358.91
    $5,358.91
    Prix promotionnel pour les nouveaux clients
    Case of 30
       $223.29
    Pallet of 720
       $5,358.91
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-345
    Fabric
    Spunbond polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    48 g/m²
  • White
    L
    39½"
    25¼"
    30¼"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $244.47
    $244.47
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 720
    Pallet of 720 - $5,867.33
    $5,867.33
    Prix promotionnel pour les nouveaux clients
    Case of 30
       $244.47
    Pallet of 720
       $5,867.33
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-344
    Fabric
    Spunbond polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    48 g/m²
  • White
    XL
    40"
    27"
    32½"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $245.25
    $245.25
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 720
    Pallet of 720 - $5,885.95
    $5,885.95
    Prix promotionnel pour les nouveaux clients
    Case of 30
       $245.25
    Pallet of 720
       $5,885.95
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-342
    Fabric
    Spunbond polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    30 g/m²
  • White
    XXL
    42¾"
    27¾"
    33"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $263.47
    $263.47
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 720
    Pallet of 720 - $6,323.33
    $6,323.33
    Prix promotionnel pour les nouveaux clients
    Case of 30
       $263.47
    Pallet of 720
       $6,323.33
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-343
    Fabric
    Spunbond polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    48 g/m²
  • White
    3XL
    43½"
    29½"
    37½"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $291.74
    $291.74
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 720
    Pallet of 720 - $7,001.86
    $7,001.86
    Prix promotionnel pour les nouveaux clients
    Case of 30
       $291.74
    Pallet of 720
       $7,001.86
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-348
    Fabric
    Spunbond polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    48 g/m²
  • White
    S
    46½"
    31½"
    39¼"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 30
    Case of 30 - $241.28
    $241.28
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 990
    Pallet of 990 - $7,962.24
    $7,962.24
    Prix promotionnel pour les nouveaux clients
    Case of 30
       $241.28
    Pallet of 990
       $7,962.24
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-346
    Fabric
    Spunbond polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    48 g/m²
48 g SPP, standard lab coat with elastic cuffs
Informations sur le produit
  • Color
    Taille de confection
    Longueur
    Chest Size
    Sleeve
    Availability
    Price
  • White
    M
    39½"
    25¼"
    30¼"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $228.68
    $228.68
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 900
    Pallet of 900 - $8,232.48
    $8,232.48
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $228.68
    Pallet of 900
       $8,232.48
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-353
    Fabric
    Spunbond polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    48 g/m²
  • White
    L
    40"
    27"
    32½"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $224.01
    $224.01
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 825
    Pallet of 825 - $7,392.26
    $7,392.26
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $224.01
    Pallet of 825
       $7,392.26
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-351
    Fabric
    Spunbond polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    48 g/m²
  • White
    XL
    42¾"
    27¾"
    33"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $224.34
    $224.34
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 900
    Pallet of 900 - $8,076.10
    $8,076.10
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $224.34
    Pallet of 900
       $8,076.10
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-350
    Fabric
    Spunbond polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    48 g/m²
  • White
    XXL
    43½"
    29½"
    37½"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $237.69
    $237.69
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 900
    Pallet of 900 - $8,556.77
    $8,556.77
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $237.69
    Pallet of 900
       $8,556.77
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-349
    Fabric
    Spunbond polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    48 g/m²
  • White
    S
    44½"
    30¾"
    38¼"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $228.15
    $228.15
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 600
    Pallet of 600 - $5,475.65
    $5,475.65
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $228.15
    Pallet of 600
       $5,475.65
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-352
    Fabric
    Spunbond polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    48 g/m²
  • White
    3XL
    44½"
    30¾"
    38¼"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $33.18
    $33.18
    Liquidation
    Case of 25
       $33.18
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
    Spécifications:
    More
    Specifications
    Cat. No.
    76311-258
    Fabric
    Spunbond polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    48 g/m²
  • White
    4XL
    46½"
    31½"
    39¼"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $255.34
    $255.34
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 750
    Pallet of 750 - $7,660.08
    $7,660.08
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $255.34
    Pallet of 750
       $7,660.08
    Spécifications:
    More
    Specifications
    Cat. No.
    414004-354
    Fabric
    Spunbond polypropylene
    Closure type
    Press studs / snaps
    Cleanroom class
    10000/ISO 7
    Pockets
    With pockets
    Gender
    Unisex
    Sleeve or leg length
    Long
    Garment application
    Cleanroom|Industrial|Laboratory
    Particle shedding (Helmke Drum)
    Level II
    Water resistance
    First line barrier
    Weight
    48 g/m²