VWR® Signature™ Lab Coats made with DuPont™ Tyvek® IsoClean® Material
Fournisseur: VWR International
Certificats
À propos de cet article
Garments provide a premium level of protection and durability while remaining comfortable
- Premium level of protection and durability while remaining comfortable
- Excellent barrier against dry particles, nonhazardous liquids, and microorganisms, designed for use in cleanroom environments
- Limited-use lab coats are antistatic treated, low linting, chemically and biologically inert
Lab coats feature serged seams, laydown collar, five-snap front, one breast pocket, two hip pockets, and open wrists. All coats feature Raglan sleeves.
Lab coats offer an excellent barrier against dry particles, nonhazardous liquids, and microorganisms, and are designed for use in cleanroom environments. Limited-use lab coats are antistatic-treated, low-linting, and chemically and biologically inert.
Spécifications
- Catalog No:
- 89128-798
- 89128-786
- 89127-192
- 89127-198
- 89125-786
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory