Chemware® Teflon® Resin Evaporating Dishes, Saint-Gobain Performance Plastics
Fournisseur: Saint Gobain Performance Plastics
Certificats
À propos de cet article
With broad pouring spouts and flat bottoms
These resin evaporating dishes, saint-gobain performance plastics are made of PTFE.
Chemically inert to virtually all materials. Heat resistant to 260°C (550°F).
Frequently Bought Together