SP Bel-Art Multipurpose Tray with Lid, Polypropylene, Bel-Art Products, a part of SP
Catalog # 74950-090
Supplier: Bel-Art Products, a Part of SP
Specifications
- Cat. no.:74950-090
- L×W×H:20.3 x 27.3 x 7.6 cm (8 x 103/4 x 3")
- Supplier No.:162300000
Specifications
About this item
Rugged, shallow tray may be used for safely storing small items, holding small vials or bottles in an ice or water bath, or holding instruments while autoclaving.
- Molded polypropylene construction
- Stackable for efficient storage
- Inside corners are smooth and rounded
- Easy to clean
Inside tray corners are smooth and rounded for easy cleaning. Stacks with identical trays and lids for efficient storage. Both tray and lid are constructed of molded polypropylene. Steam autoclavable at 121°C (250°F).