PYREX® Reducing Adapters with [ST] Joints, Corning
 :   Corning
PYREX® Reducing Adapters with [ST] Joints, Corning
 :  Corning
PYREX® Reducing Adapters with [ST] Joints, Corning
PYREX® Reducing Adapters with [ST] Joints, Corning
PYREX® glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Expansion adapter from small cone to large socket.
  • Catalog No:
  • 89091-642
  • 89091-644
  • 62900-088
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
Product Family Options
  • Longueur
    Bottom Inner [ST] Joint Size
    Top Outer ST Joint Size
  • 105 mm
    19/38
    [ST] 24/40
    Case of 6
    Case of 6 - $809.00
    $809.00
    Case of 6
       $809.00
     : 8800-2419
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    89091-642
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 105mm
    24/40
    [ST] 29/42
    Case of 6
    Case of 6 - $876.00
    $876.00
    Case of 6
       $876.00
     : 8800-2924
      Low Manufacturing Impact
    Specifications
    Cat. No.
    89091-644
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
      Low Manufacturing Impact
  • 110mm
    24/40
    [ST] 34/45
    Case of 1
    Case of 1 - $180.00
    $180.00
    Case of 1
       $180.00
     : 8800-3424
      Low Manufacturing Impact
    Specifications
    Cat. No.
    62900-088
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
      Low Manufacturing Impact