Synthware Reducing Adapters, Kemtech America
Supplier: Kemtech America
Jump to:
À propos de cet article
Reducing adapters are used for connecting dissimilar [ST] joints.
- Top outer [ST] joint
- Bottom inner [ST] joint
Features an outer [ST] joint at the top and inner [ST] joint at the bottom.
7 Options Available Below
Product Details & Documents
Reducing adapters are used for connecting dissimilar [ST] joints.
- Top outer [ST] joint
- Bottom inner [ST] joint
Features an outer [ST] joint at the top and inner [ST] joint at the bottom.
Product Family Options
Informations sur le produit
- Top ST Joint SizeBottom [ST] Joint SizeAvailabilityPrice
- Supplier Number: A201024
- Supplier Number: A201419
- Supplier Number: A201424
- Supplier Number: A201924
- Supplier Number: A202429
- Supplier Number: A202434
- Supplier Number: A202445