SCOTT® Hard Roll Paper Towels, Kimberly-Clark Professional®
Catalog # CA89004-434
Supplier: Kimberly-Clark
Specifications
- Width:20.3 cm (8")
- Cat. no.:CA89004-434
- Roll Length:244 m (800')
- Supplier No.:01040
Specifications
About this item
Made with Absorbency Pockets® for better absorbency and softness. Color: white.