Synthware Distillation Connecting Adapters
Supplier: Kemtech America
Jump to:
À propos de cet article
Use distillation connecting adapter as a component in atmospheric or vacuum distallations.
- Inner joints at bottom and side arm
- Outer joint at top
The lower end and side arm have [ST] inner joints. Upper end has [ST] outer joint.
8 Options Available Below
Product Details & Documents
Use distillation connecting adapter as a component in atmospheric or vacuum distallations.
- Inner joints at bottom and side arm
- Outer joint at top
The lower end and side arm have [ST] inner joints. Upper end has [ST] outer joint.
Product Family Options
Informations sur le produit
- Bottom [ST] Joint SizeTop ST Joint SizeSide ST Joint SizeAvailabilityPrice
- Supplier Number: A351014
- Supplier Number: A351420
- Supplier Number: A351019
- Supplier Number: A351922
- Supplier Number: A351024
- Supplier Number: A352440
- Supplier Number: A351424
- Supplier Number: A354914