Nalgene® Sterilizing Pans, Polypropylene, Thermo Scientific
Nalgene® Sterilizing Pans, Polypropylene, Thermo Scientific
Nalgene® Sterilizing Pans, Polypropylene, Thermo Scientific
Certificates
Nalgene® Sterilizing Pans, Polypropylene, Thermo Scientific
Nalgene® Sterilizing Pans, Polypropylene, Thermo Scientific
2 Options Available Below
Product Details & Documents
Useful for drying and sterilizing instruments and labware, and for collecting and washing soiled lab equipment
Rolled edges for extra stiffness. Handles for easy carrying. Tapered for nesting. Autoclavable.
Documents
Product Family Options
Product Information
  • L×W×H
    Availability
    Price
  • 36 x 31 x 14 cm (143/16 x 127/32 x 51/2")
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $97.18
    $97.18
    New Customer Promotional Price
    Log in for availability
    Case of 6
    Case of 6 - $583.08
    $583.08
    New Customer Promotional Price
    Each
       $97.18
    Case of 6
       $583.08
    Supplier Number: 6900-0010
  • 54.3 x 43.5 x 13 cm (213/8 x 171/8 x 51/8")
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $171.95
    $171.95
    New Customer Promotional Price
    Log in for availability
    Case of 6
    Case of 6 - $1,031.70
    $1,031.70
    New Customer Promotional Price
    Each
       $171.95
    Case of 6
       $1,031.70
    Supplier Number: 6900-0020