Spill Containment Pallet/Platforms with Grating, Eagle Manufacturing
Supplier:   Eagle Manufacturing
Spill Containment Pallet/Platforms with Grating, Eagle Manufacturing
Certificates
Spill Containment Pallet/Platforms with Grating, Eagle Manufacturing
Spill Containment Pallet/Platforms with Grating, Eagle Manufacturing
4 Options Available Below
Product Details & Documents
Low-profile pallet and platform units support 208.2L (55gal.) drums and provide secondary spill containment.
  • Low-profile units support 208.2L (55gal.) drums
  • Provide secondary spill containment
All-polyethylene construction with excellent chemical resistance. Translucent tub is safety yellow with black grating supports. Grating has 4.4cm (13/4") square openings and removes easily for cleaning. Pallet unit 56617-333 is forkliftable and has a 1.9cm (3/4") drainplug. Ramp 56610-212 fits platform units.
 : Meet EPA requirement 40 CFR 264.175.
Product Family Options
Informations sur le produit
  • Désignation
    Hauteur
    Longueur
    Load Capacity
    Width
    Sump Capacity
    Weight
    Availability
    Price
  • Six-Drum Platform
    6½"
    77"
    8000 lbs.
    51½"
    61 gal.
    88 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,483.41
    $1,483.41
    Each
       $1,483.41
    Supplier Number: 1686
  • Eight-Drum Platform
    6½"
    102"
    10000 lbs.
    51½"
    82 gal.
    116 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,026.23
    $2,026.23
    Each
       $2,026.23
    Supplier Number: 1688
  • Four-Drum Platform, In-Line
    6¾"
    106"
    8000 lbs.
    30¼"
    63 gal.
    74 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,100.57
    $1,100.57
    Each
       $1,100.57
    Supplier Number: 1647
  • Four-Drum Pallet
    10"
    51½"
    8000 lbs.
    51½"
    62 gal.
    65 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,460.02
    $2,460.02
    Each
       $2,460.02
    Supplier Number: 1645
Accessories
Informations sur le produit
  • Catalog #
    L×W×H
    Load Capacity
    Availability
    Price
  • Ramp
    Ramp
    56610-212
    115.5 x 81.2 x 20.3 cm (451/2 x 32 x 8")
    680 kg (1500 lbs.)
    Log in pour la disponibilité
    Each - $1,347.78