Double Valve Oxygen Bomb, Parr®
Designed to be easy to handle, easy to maintain and safe for determination of heats of combustion in environmental, energy and nutrition research
Provides rapid and straightforward combustion, simultaneously releasing up to 8 Kcal while oxidizing individual chemical elements for recovery analysis. Manufactured of columbium (niobium) stabilized steel (Carpenter No. 20 Cb-3), the 1108 bomb offers excellent resistance to generated nitric and sulfuric acids. The high strength bronze screw cap slides smoothly without seizing.
Used in Parr® Oxygen Bomb Calorimeters.
Max. charging pressure: 40 atm
: Factory tested according to ANSI/ASTM Method 144-64(1976), “Recommended Practice for Safe Use of Oxygen Combustion Bombs.”
: Supplied with ANSI/ASTM certificate. Ignition units and the oxygen filling connection must be ordered separately.
- Catalog No:
- 19422-051
- 19440-020
- Dimensions:
- 16.8 H x 9.5 O.D. cm
- Int. volume:
- 340 mL
- Weight:
- 29.4 kg (64.8 lbs.)
Frequently Bought Together