Synthware Drying Tube Adapters, Straight, Kemtech America
Fournisseur: Kemtech America
Certificats
À propos de cet article
Drying tube adapters are used for keeping a reaction vessel free of moisture.
- Straight
- With top and bottom joints
Adapters have both a top and bottom joint.
Frequently Bought Together