PYREX® Drying Trays, Corning
PYREX® Drying Trays, Corning
Supplier:   Corning
PYREX® Drying Trays, Corning
Supplier:  Corning
Certificates
PYREX® Drying Trays, Corning
PYREX® Drying Trays, Corning
3 Options Available Below
Product Details & Documents
These dishes made of PYREX® borosilicate glass will withstand hot air or steam sterilization and are useful for drying, staining or developing trays.
  • Excellent chemical resistance
  • Good temperature resistance
  • Easy to clean
They can be used with gel electrophoresis or paper chromatography.
Caution: For oven use only. Do not use over direct heat.
Product Family Options
Informations sur le produit
  • Volume
    L×W×H
    Availability
    Price
  • 1750 ml
    27.2 x 18 x 5 cm (1023/32 x 7 x 2")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 4
    Case of 4 - $139.12
    $139.12
    Prix promotionnel pour les nouveaux clients
    Case of 4
       $139.12
    Supplier Number: 3175-7
  • 2000 ml
    20 x 20 x 5.5 cm (8 x 8 x 25/32")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 4
    Case of 4 - $108.99
    $108.99
    Prix promotionnel pour les nouveaux clients
    Case of 4
       $108.99
    Supplier Number: 3175-8
  • 3500 ml
    34.9 x 24.9 x 6 cm (1323/32 x 913/16 x 211/32")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 4
    Case of 4 - $226.00
    $226.00
    Case of 4
       $226.00
    Supplier Number: 3175-10