Mouse MAdCAM1 (Mucosal Addressin Cell Adhesion Molecule 1) ELISA Kit
Catalogue # 77518-020
Fournisseur: AFG Bioscience
Numéro CAS:
Spécifications
- Assay Type:Sandwich
- Host:Mouse
- Target protein:MAdCAM1 (Mucosal Addressin Cell Adhesion Molecule 1)
- Size:96 wells
- Sample type:Serum, plasma, tissue homogenates and other biological fluids
- Time to Results:3.5 h
- Shelf life:6 months (4 °C); 12 months (–20 °C)
- Detection range:0.16 - 10 ng/ml
- Storage temperature:2…8 °C
- Application:CD and adhesion molecule
- Sensitivity:0.059 ng/ml
- Regulatory status:RUO
- Cat. no.:77518-020