Cubis® II Semi-Micro Balances
Semi-micro balances come with a readability of 0.01 mg or 10 µg and a maximum weighing capacity of up to 220 g.
- Superior weighing performance: Fast measurement time and accurate weighing results are guaranteed thanks to next generation monolithic weighing system and the engineering solution compensating for environmental changes
- No more issues with charged samples: Glass draft shields coated with conductive layer prevent outer electrostatic effects and the built-in ionizer effectively eliminates electrostatic charges from samples
- Cleaning process guidance: Intuitive process for daily and advanced cleaning is now available in all Cubis® II MCA. The cleaning QApp provides visual guidance, information about chemical compatibility as well as electronical tracking of these events
- Hardware upgradeability: Hardware features like motorized draft shield function or built-in ionizer can be activated post-purchase. An automated inner draft shield is available as a klick-in accessory which is recommended for use under challenging environmental conditions (e.g. in drafty environments)
- Compliance and Data Integrity: The end-to-end data integrity, technical controls for 21 CFR Part 11 compliance, integrated audit-trail and state-of-the-art user management ensure laboratories can meet regulatory requirements. These features are directly available on the Cubis® II balances, without the need of additional software
- Balances Fleet Management: The Ingenix Suite is a flexible, open solution that works with or without an ELN/LIM system. It offers unlimited connections to easily manage the entire Cubis® II MCA lab balance fleet across all labs within the same network
- Ease of use: The teaching function with learning capability of the motorized draft shield, guided workflows for many different applications (QApps), automated motorized leveling and automatic internal adjustment (isoCAL) help with easy and error-free operation of the balance
Semi-micro balances are used for weighing small sample amounts down to the µg-range. To meet the requirements of your specific application, specially designed weighing pans, sample holders and software applications are available to simplify your weighing processes and laboratory workflows.
- Catalog No:
- CA77476-512
- CA77476-510
- CA77476-520
- CA77476-492
- CA77476-518
- CA77476-516
- CA77476-514
- CA77476-548
- CA77476-546
- CA77476-544
- CA77476-490
- CA77476-488
- CA77476-558
- CA77476-556
- CA77476-554
- CA77476-552
- CA77476-550
- CA77476-508
- CA77476-528
- CA77476-542
- CA77476-540
- CA77476-538
- CA77476-536
- CA77476-534
- CA77476-532
- CA77476-530
- CA77476-500
- CA77476-526
- CA77476-498
- CA77476-524
- CA77476-496
- CA77476-494
- CA77476-522
- CA77476-506
- CA77476-504
- CA77476-502
- Operating temperature range:
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- Plug type:
- IEC plug
- IEC plug
- IEC plug
- IEC plug
- IEC plug
- IEC plug
- IEC plug
- IEC plug
- IEC plug
- IEC plug
- IEC plug
- IEC plug
- IEC plug
- IEC plug
- IEC plug
- IEC plug
- IEC plug
- IEC plug
- IEC plug
- IEC plug
- IEC plug
- IEC plug
- IEC plug
- IEC plug
- IEC plug
- IEC plug
- IEC plug
- IEC plug
- IEC plug
- IEC plug
- IEC plug
- IEC plug
- IEC plug
- IEC plug
- IEC plug
- IEC plug
- Communication interface:
- USB-A;USB-B;USB-C Interface
- USB-A;USB-B;USB-C Interface
- USB-A;USB-B;USB-C Interface
- USB-A;USB-B;USB-C Interface
- USB-A;USB-B;USB-C Interface
- USB-A;USB-B;USB-C Interface
- USB-A;USB-B;USB-C Interface
- USB-A;USB-B;USB-C Interface
- USB-A;USB-B;USB-C Interface
- USB-A;USB-B;USB-C Interface
- USB-A;USB-B;USB-C Interface
- USB-A;USB-B;USB-C Interface
- USB-A;USB-B;USB-C Interface
- USB-A;USB-B;USB-C Interface
- USB-A;USB-B;USB-C Interface
- USB-A;USB-B;USB-C Interface
- USB-A;USB-B;USB-C Interface
- USB-A;USB-B;USB-C Interface
- USB-A;USB-B;USB-C Interface
- USB-A;USB-B;USB-C Interface
- USB-A;USB-B;USB-C Interface
- USB-A;USB-B;USB-C Interface
- USB-A;USB-B;USB-C Interface
- USB-A;USB-B;USB-C Interface
- USB-A;USB-B;USB-C Interface
- USB-A;USB-B;USB-C Interface
- USB-A;USB-B;USB-C Interface
- USB-A;USB-B;USB-C Interface
- USB-A;USB-B;USB-C Interface
- USB-A;USB-B;USB-C Interface
- USB-A;USB-B;USB-C Interface
- USB-A;USB-B;USB-C Interface
- USB-A;USB-B;USB-C Interface
- USB-A;USB-B;USB-C Interface
- USB-A;USB-B;USB-C Interface
- USB-A;USB-B;USB-C Interface
- Voltage:
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
Frequently Bought Together