PinPoint™ Pipette Controller
PinPoint™ Pipette Controller
PinPoint™ Pipette Controller
Certificates
PinPoint™ Pipette Controller
PinPoint™ Pipette Controller
5 Options Available Below
Product Details & Documents
Digital display makes transferring liquids easier.
  • Compatible with glass or plastic serological pipettes (0.1 to 100 ml)
  • LCD indicates battery charge and 6 speed levels
  • Fills a 25 ml pipette in less than 5 seconds
  • Lithium-ion battery charges fully within 3 hours and can provide up to 8 hours of use
Ultra-light and powerful pipette controller is designed for comfort as well as accuracy. Intuitive 6 level speed setting and sensitive triggers provide a wide range of aspirating/dispensing speeds. Pipette controller is ergonomically designed for extended usage.
Certifications: Meets the requirements of the applicable EU directives, UK regulations, UL, and CAN/CSA.
Ordering information: The PinPoint™ Pipette Controller comes with an AC adapter with USB connection, a 0.45 μm hydrophobic filter, a user manual, wall mount, and two-position desk stand.
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • PinPoint™ pipette controller
    Log in for availability
    Log in for availability
    Each
    Each - $317.55
    $317.55
    New Customer Promotional Price
    Each
       $317.55
    Supplier Number: UNPINPNT01
  • PinPoint™ silicone adapter
    Log in for availability
    Log in for availability
    Each
    Each - $7.08
    $7.08
    New Customer Promotional Price
    Each
       $7.08
    Supplier Number: UNPINPNT02
  • PinPoint™ nosecone
    Log in for availability
    Log in for availability
    Each
    Each - $10.03
    $10.03
    New Customer Promotional Price
    Each
       $10.03
    Supplier Number: UNPINPNT03
  • PinPoint™ 0.22 μm filters
    Log in for availability
    Log in for availability
    Pack of 5
    Pack of 5 - $48.66
    $48.66
    New Customer Promotional Price
    Pack of 5
       $48.66
    Supplier Number: UNPINPNT05
  • PinPoint™ 0.45 μm filters
    Log in for availability
    Log in for availability
    Pack of 5
    Pack of 5 - $48.66
    $48.66
    New Customer Promotional Price
    Pack of 5
       $48.66
    Supplier Number: UNPINPNT04