Engino® GINOBOT Basic Edition
Engino® GINOBOT Basic Edition
Engino GINOBOT basic edition
Catalog # 470356-040
Supplier #:  UNE491
Engino® GINOBOT Basic Edition
Catalog # 470356-040
Engino® GINOBOT Basic Edition
Engino® GINOBOT Basic Edition
Engino GINOBOT basic edition
Price
280,80 $
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Kit of 1
Product Details & Documents
Take coding and programming to the next level in a fun and exciting way.
  • Download easy-to-follow instructions, theory and quizzes
  • Includes sample programs
  • Online step by step building guide
  • Encourages creativity and critical thinking
  • Versatile with the ability to build different models
GinoBot™ is a highly programmable robot ready to be used straight out of the box. Developed by a high-level research team of engineers and academics, GinoBot™ is a unique tool for teaching STEM disciplines, computational thinking and digital literacy with fun and hands-on experiential activities. The body of GinoBot™ has numerous built-in ENGINO® connecting geometries so that students can expand their robot with structural and mechanical parts, making more advanced models.
Teachers can follow STEM interdisciplinary activities and apply inquiry-based learning methodologies to engage and inspire their students. The vehicle’s powerful processor connects with USB, bluetooth and Wi-Fi and can be programmed with KEIRO, but also Python and C++ once a Micro:bit or Arduino are installed. The 4-wheeled robot can have its tires converted to a cat-truck and is surrounded by three proximity sensors. It has two floor color sensors and two encoder motors for precise motion.
Suggested for students working in groups of 3 or 4.
Delivery information: Kit includes 16 plastic parts, 87 connecting points.
Spécifications
  • Désignation:
    Engino GINOBOT basic edition