ContactUsLinkComponent
Engino® STEM and Robotics Mini Set
Catalogue # 470356-026
undefined
Engino® STEM and Robotics Mini Set
Catalogue # 470356-026
Supplier Number:  UNE201
Spécifications
  • Description:
    Engino STEM and robotics mini set
  • Cat. no.:
    470356-026
Spécifications
À propos de cet article
Introduce coding and programming in a fun an easy way.
  • Download easy-to-follow instructions, theory and quizzes
  • Includes sample programs
  • Online step by step building guide
  • Encourages creativity and critical thinking
  • Versatile with the ability to build different models
The STEM and Robotics ERP Mini education set is specially designed for early primary school students of ages 7 to 9. It combines the core subjects of STEM together with Robotics. The set comes in a convenient plastic storage tub that contains a large number of Engino - Qboidz plastic parts along with robotic devices such as controller Mini, DC motor, LED, touch sensor, IR sensor.
The extra ENGINO® technical parts allow the construction of 40 complex models that come with theory and experimental activities to cover not only engineering design but also Coding, Mechanics and Science. There are two generations of robotic models doable with this set. Simpler models using the Qboidz parts which aim to introduce coding to the younger students, and complex ones with the classic ENGINO parts for more advanced models.
Suggested for students working in groups of 2 or 3.
Ordering information: Required but not included: (3) AAA batteries.
Delivery information: Kit includes 312 plastic parts, 1596 connecting points, battery compartment for 3×AAA batteries, mini controller with four input-output ports, connectivity via USB and bluetooth, onboard membrane buttons for manual programming, two IR sensors and one touch sensor, two DC motors, one LED, red, plastic tub (43×31×15 cm) with internal dividers.