ContactUsLinkComponent
Engino® STEM Newton's Laws and Inclined Planes
Engino® STEM Newton's Laws and Inclined Planes
Catalog # 470356-056
undefined
Engino® STEM Newton's Laws and Inclined Planes
Catalog # 470356-056
Supplier Number:  STEM902
Specifications
  • Description:
    Engino STEM newton's laws and inclined planes
  • Cat. no.:
    470356-056
Specifications
About this item
Learn about Newton's laws and design structures to experiment with potential and kinetic energy.
  • Includes a manual with instructions, theory and quizzes
  • Online step by step building guide
  • Encourages creativity and critical thinking
  • Versatile with the ability to build different models
Learn all about Newton's laws of motion which are the basis of classical mechanics that still describe most everyday life situations. Experiment with kinetic and potential energy in order to discover the properties of energy and how it is transformed from one form to the other. Build six working models such as a crash test rig, a rocket launcher, a sharpening wheel, a bow and arrow, a rubber car and an inertia test platform.
You can find easy-to-follow building instructions for all models either online or in the booklet included. The booklet provides detailed explanations of the different scientific principles applied and incorporates innovative experimental activities for hands-on learning. A Quiz section is also available to challenge your newly acquired knowledge!